Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

33-0550

Sigma-Aldrich

Cyanocobalamin

SAJ special grade, ≥95.0%

Synonym(s):

Vitamin B12, α-(5,6-Dimethylbenzimidazolyl)cyanocobamide, CN-Cbl, Cyanocob(III)alamin, Cyanocobalamin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C63H88CoN14O14P
CAS Number:
Molecular Weight:
1355.37
Beilstein/REAXYS Number:
4122889
UNSPSC Code:
41100000
PubChem Substance ID:
Pricing and availability is not currently available.

grade

SAJ special grade

assay

≥95.0%

form

solid

availability

available only in Japan

color

white

mp

>300 °C (decomposes before melting)

storage temp.

2-8°C

SMILES string

C/C(C1=N[C@]([C@]([C@H](CC(N)=O)[C@@]/2(C)CCC(NC[C@@H](OP([O-])(O[C@H]3[C@@H](O)[C@@H](N4C=[N]5C6=C4C=C(C)C(C)=C6)O[C@@H]3CO)=O)C)=O)([H])N([Co+]5C#N)C2=C(C)/C([C@@H](CCC(N)=O)C/7(C)C)=NC7=C/8)(C)[C@@](C)(CC(N)=O)[C@@H]1CCC(N)=O)=C9N=C8[C@@H](CCC(N)=O)[C@

InChI

1S/C62H90N13O14P.CN.Co/c1-29-20-39-40(21-30(29)2)75(28-70-39)57-52(84)53(41(27-76)87-57)89-90(85,86)88-31(3)26-69-49(83)18-19-59(8)37(22-46(66)80)56-62(11)61(10,25-48(68)82)36(14-17-45(65)79)51(74-62)33(5)55-60(9,24-47(67)81)34(12-15-43(63)77)38(71-55)23-42-58(6,7)35(13-16-44(64)78)50(72-42)32(4)54(59)73-56;1-2;/h20-21,23,28,31,34-37,41,52-53,56-57,76,84H,12-19,22,24-27H2,1-11H3,(H15,63,64,65,66,67,68,69,71,72,73,74,77,78,79,80,81,82,83,85,86);;/q;;+2/p-2/t31-,34-,35-,36-,37+,41-,52-,53-,56-,57?,59-,60+,61+,62+;;/m1../s1

InChI key

RMRCNWBMXRMIRW-QJRSUKKJSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sabina Lutz et al.
Journal of the American Animal Hospital Association, 49(3), 197-203 (2013-03-29)
Juvenile cobalamin deficiency is a rare disease in border collies and its diagnosis requires a high level of clinical suspicion. The goal of this study was to increase awareness of this disease by describing the clinical and laboratory findings in
David Preiss et al.
The lancet. Diabetes & endocrinology, 2(2), 116-124 (2014-03-14)
Metformin reduces cardiovascular risk in patients with type 2 diabetes seemingly independent of lowering blood glucose concentration. We assessed the cardiovascular effects of metformin in individuals without type 2 diabetes. We did a single-centre, double-blind, placebo-controlled trial at the Glasgow
Bamini Gopinath et al.
The American journal of clinical nutrition, 98(1), 129-135 (2013-05-03)
Epidemiologic evidence of a relation between serum total homocysteine (tHcy), vitamin B-12, and folate and age-related macular degeneration (AMD) is inconsistent and unresolved. In this cohort study, we aimed to investigate associations between intakes and serum concentrations of folate and
Johan Frederik Berg Arendt et al.
Journal of the National Cancer Institute, 105(23), 1799-1805 (2013-11-20)
A substantial proportion of patients referred for plasma vitamin B12 (cobalamin [Cbl]) measurement present with high Cbl levels, which have been reported in patients with different cancer types. However, the cancer risk among patients with newly diagnosed high Cbl levels
Bert-Ram Sah et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 55(1), 43-49 (2013-12-18)
Targeting cancer cells with vitamin B12 (cobalamin) is hampered by unwanted physiologic tissue uptake mediated by transcobalamin. Adhering to good manufacturing practice, we have developed a new (99m)Tc-cobalamin derivative ((99m)Tc(CO)3-[(4-amido-butyl)-pyridin-2-yl-methyl-amino-acetato] cobalamin, (99m)Tc-PAMA-cobalamin). The derivative shows no binding to transcobalamin but

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service