Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

33-0540

Sigma-Aldrich

Pyridoxine hydrochloride

SAJ special grade, ≥98.0%

Synonym(s):

Adermine hydrochloride, PN HCl, Pyridoxol hydrochloride, Vitamin B6 hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H11NO3 · HCl
CAS Number:
Molecular Weight:
205.64
Beilstein/REAXYS Number:
3632435
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
Pricing and availability is not currently available.

grade

SAJ special grade

assay

≥98.0%

form

powder

availability

available only in Japan

color

white

mp

214-215 °C (lit.)

storage temp.

20-25°C

SMILES string

Cl[H].Cc1ncc(CO)c(CO)c1O

InChI

1S/C8H11NO3.ClH/c1-5-8(12)7(4-11)6(3-10)2-9-5;/h2,10-12H,3-4H2,1H3;1H

InChI key

ZUFQODAHGAHPFQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vijay P Singh et al.
The Journal of organic chemistry, 78(4), 1478-1487 (2013-01-16)
Vitamin B6 is involved in a variety of enzymatic transformations. Some recent findings also indicate an antioxidant role of the vitamin in biological systems. We set out to turn pyridoxine (1a) into a catalytic chain-breaking and hydroperoxide-decomposing antioxidant by replacing
Manfred Eggersdorfer et al.
Angewandte Chemie (International ed. in English), 51(52), 12960-12990 (2012-12-05)
The discovery of vitamins as essential factors in the diet was a scientific breakthrough that changed the world. Diseases such as scurvy, rickets, beriberi, and pellagra were recognized to be curable with an adequate diet. These diseases had been prevalent
Liat Ashkenazi-Hoffnung et al.
The Israel Medical Association journal : IMAJ, 15(1), 23-26 (2013-03-15)
Diclectin (pyridoxine 10 mg and doxylamine 10 mg) has traditionally been used to treat nausea and vomiting of pregnancy (NVP); however, this drug is unavailable in many countries. To evaluate the efficacy and safety of a simple bi-daily treatment regimen
Barbara Plecko et al.
Neurology, 82(16), 1425-1433 (2014-03-25)
To determine whether patients with pyridoxine-responsive seizures but normal biomarkers for antiquitin deficiency and normal sequencing of the ALDH7A1 gene may have PNPO mutations. We sequenced the PNPO gene in 31 patients who fulfilled the above-mentioned criteria. We were able
Ilan Matok et al.
Journal of clinical pharmacology, 53(3), 334-338 (2013-02-28)
Although Diclectin (doxylamine/pyridoxine delayed-released combination) is widely used in Canada, its pharmacokinetics (PK) during pregnancy has never been described. The objective of this study was to compare the PK of doxylamine/pyridoxine delayed-released combination in pregnant versus nonpregnant women. The apparent

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service