Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

33-0510

Sigma-Aldrich

Thiamine hydrochloride

SAJ special grade, ≥98.5%

Synonym(s):

Aneurine hydrochloride, Vitamin B1 hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H17ClN4OS · HCl
CAS Number:
Molecular Weight:
337.27
Beilstein/REAXYS Number:
3851771
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
Pricing and availability is not currently available.

grade

SAJ special grade

assay

≥98.5%

form

powder

availability

available only in Japan

color

white

mp

250 °C (dec.) (lit.)

storage temp.

20-25°C

SMILES string

CC1=NC(N)=C(C[N+]2=CSC(CCO)=C2C)C=N1.Cl.[Cl-]

InChI

1S/C12H17N4OS.2ClH/c1-8-11(3-4-17)18-7-16(8)6-10-5-14-9(2)15-12(10)13;;/h5,7,17H,3-4,6H2,1-2H3,(H2,13,14,15);2*1H/q+1;;/p-1

InChI key

DPJRMOMPQZCRJU-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joseph E Kerschner et al.
Archives of otolaryngology--head & neck surgery, 135(1), 33-39 (2009-01-21)
To investigate genetic differences in middle ear mucosa (MEM) with nontypeable Haemophilus influenzae (NTHi) infection. Genetic upregulation and downregulation occurs in MEM during otitis media (OM) pathogenesis. A comprehensive assessment of these genetic differences using the techniques of complementary DNA
Samir Ahmed El-Shazly et al.
International journal of immunopathology and pharmacology, 28(2), 227-239 (2015-03-31)
Diabetes mellitus (DM) is a chronic metabolic disorder characterized by hyperglycemia due to abnormalities in either insulin secretion or action. A range of vanadium complexes have been synthesized and demonstrated to be effective in lowering hyperglycemia. Thiamine administration was also
Ashish K Singh et al.
Alcoholism, clinical and experimental research, 38(7), 2127-2137 (2014-06-17)
Chronic ethanol (EtOH) consumption is associated with oxidative tissue damage, decrease in antioxidant enzyme activities, and increase in hepatic and plasma lipids. This study investigates the effect of modified probiotic Escherichia coli Nissle 1917 (EcN) secreting pyrroloquinoline quinone (PQQ) against
Kris Niño G Valdehuesa et al.
Bioprocess and biosystems engineering, 38(9), 1761-1772 (2015-06-07)
Biosynthetic pathways for the production of biofuels often rely on inherent aldehyde reductases (ALRs) of the microbial host. These native ALRs play vital roles in the success of the microbial production of 1,3-propanediol, 1,4-butanediol, and isobutanol. In the present study
Katherine R Heal et al.
Rapid communications in mass spectrometry : RCM, 28(22), 2398-2404 (2014-10-11)
Vitamin B(12) is an essential nutrient for more than half of surveyed marine algae species, but methods for directly measuring this important cofactor in seawater are limited. Current mass spectrometry methods do not quantify all forms of B(12), potentially missing

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service