Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

33-0450

Sigma-Aldrich

1-Vinyl-2-pyrrolidinone

SAJ first grade, ≥99.0%

Synonym(s):

1-Vinyl-2-pyrrolidone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H9NO
CAS Number:
Molecular Weight:
111.14
Beilstein/REAXYS Number:
110513
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:
grade:
SAJ first grade
assay:
≥99.0%
bp:
92-95 °C/11 mmHg (lit.)
vapor pressure:
0.1 mmHg ( 24 °C)
Pricing and availability is not currently available.

grade

SAJ first grade

vapor density

3.8 (vs air)

vapor pressure

0.1 mmHg ( 24 °C)

assay

≥99.0%

form

liquid

autoignition temp.

685 °F

contains

~25 ppm N,N′-di-sec-butyl-p-phenylenediamine as inhibitor

expl. lim.

10 %

availability

available only in Japan

dilution

(for analytical testing)

refractive index

n20/D 1.512 (lit.)

bp

92-95 °C/11 mmHg (lit.)

density

1.04 g/mL at 25 °C (lit.)

SMILES string

C=CN1CCCC1=O

InChI

1S/C6H9NO/c1-2-7-5-3-4-6(7)8/h2H,1,3-5H2

InChI key

WHNWPMSKXPGLAX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - STOT RE 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

213.8 °F

flash_point_c

101 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhaoqiang Wu et al.
Colloids and surfaces. B, Biointerfaces, 96, 37-43 (2012-04-19)
A new method for the modification of poly(dimethylsiloxane) (PDMS) elastomer surfaces with hydrophilic poly(N-vinylpyrrolidone) (PVP) has been developed. PVP chains were grafted from the PDMS surface by surface-initiated atom transfer radical polymerization (SI-ATRP). The resulting surfaces were characterized by X-ray
Kris N J Stevens et al.
Biomaterials, 32(5), 1264-1269 (2010-11-26)
Central venous catheters (CVCs) have become indispensable in the treatment of neonates and patients undergoing chemotherapy or hemodialysis. A CVC provides easy access to the patient's circulation, thus enabling facile monitoring of hemodynamic parameters, nutritional support, or administration of (cytostatic)
Andrew M Telford et al.
Langmuir : the ACS journal of surfaces and colloids, 27(23), 14207-14217 (2011-10-21)
We investigated the dewetting of metastable poly(N-vinylpyrrolidone) (PNVP) thin films (45 nm) on top of polystyrene (PS) thin films (58 nm) as a function of annealing temperature and molecular weight of PS (96 and 6850 kg/mol). We focused on the
Ana C Borges et al.
Acta biomaterialia, 7(9), 3412-3421 (2011-06-10)
The swelling and compressive mechanical behavior as well as the morphology and biocompatibility of composite hydrogels based on Tween® 20 trimethacrylate (T3), N-vinyl-2-pyrrolidone (NVP) and nanofibrillated cellulose (NFC) were assessed in the present study. The chemical structure of T3 was
Hiroshi Ohno et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 16(34), 10409-10417 (2010-07-14)
Recombinant antifreeze proteins (AFPs), representing a range of activities with respect to ice growth inhibition, were investigated for their abilities to control the crystal formation and growth of hydrocarbon hydrates. Three different AFPs were compared with two synthetic commercial inhibitors

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service