Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

33-0295

Sigma-Aldrich

Petrolatum Jelly

SAJ first grade, white

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41100000

grade

SAJ first grade

availability

available only in Japan

color

white

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vaseline is a registered trademark of Unilever Supply Chain, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katrin Lorenz-Baath et al.
ChemMedChem, 7(8), 1490-1495 (2012-07-13)
Bacterial infections of skin and soft tissue represent a major health threat, especially if they are caused by multidrug-resistant strains such as MRSA. Novel treatment options for topical application are urgently needed, and even if new drug candidates are identified
Chelsea Pinnix et al.
International journal of radiation oncology, biology, physics, 83(4), 1089-1094 (2011-12-17)
To determine the efficacy of an emulsion containing hyaluronic acid to reduce the development of ≥ Grade 2 radiation dermatitis after adjuvant breast radiation compared with best supportive care. Women with breast cancer who had undergone lumpectomy and were to
Ask the doctor. I know someone who swears by Vaseline as a face cream. What do you think?
Kenneth Arndt
Harvard health letter, 37(6), 8-8 (2012-05-04)
P Mezzana et al.
The Journal of laryngology and otology, 125(10), 1073-1074 (2011-08-03)
The authors discuss a case of conjunctival irritation and epiphora in a patient who had undergone dacryocystorhinostomy three years earlier. Case report, and a review of the world literature concerning lacrimal canalicular air regurgitation after dacryocystorhinostomy. A 70-year-old woman, who
Luis Gorospe et al.
Clinical imaging, 37(1), 163-166 (2012-12-05)
A 67-year-old female ex-smoker was referred to our hospital after chest radiographs revealed a nonresolving opacity in the right lower lung. Her past medical history was significant for laryngeal cancer. A whole-body positron emission tomography/computed tomography (CT) confirmed an ill-defined

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service