Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

30-5150

Sigma-Aldrich

Triton X-405, reduced

CP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C8H17)C6H4(OCH2CH2)n OH
UNSPSC Code:
12161900
Pricing and availability is not currently available.

grade

CP

form

liquid

availability

available only in Japan

technique(s)

blocking: suitable
cell culture | mammalian: suitable
protein purification: suitable

Legal Information

Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

483.8 °F - closed cup

flash_point_c

251 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service