Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

30-4640

Sigma-Aldrich

Trimethyl phosphate

SAJ first grade, ≥95.0%

Synonym(s):

TMP, TMPA, TMPO

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)3PO
CAS Number:
Molecular Weight:
140.07
Beilstein/REAXYS Number:
1071731
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
assay:
≥95.0%
grade:
SAJ first grade
form:
liquid
Pricing and availability is not currently available.

grade

SAJ first grade

assay

≥95.0%

form

liquid

availability

available only in Japan

refractive index

n20/D 1.395 (lit.)

bp

197 °C (lit.)

mp

−46 °C (lit.)

density

1.197 g/mL at 25 °C (lit.)

storage temp.

15-25°C

SMILES string

COP(=O)(OC)OC

InChI

1S/C3H9O4P/c1-5-8(4,6-2)7-3/h1-3H3

InChI key

WVLBCYQITXONBZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Muta. 1B - Skin Irrit. 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 1

flash_point_f

224.6 °F

flash_point_c

107 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel Z Brunengraber et al.
Analytical biochemistry, 306(2), 278-282 (2002-07-19)
We have developed an assay for determining the 18O enrichment of water in biological fluids. Urine, plasma, or whole blood is reacted with phosphorous pentachloride to yield phosphoric acid. Derivatization of phosphoric acid with diazomethane generates trimethyl phosphate. The enrichment
R V Banerjee et al.
Biochemistry, 27(25), 9062-9070 (1988-12-13)
The transfer of 17O and/or 18O from (COOH-17O or -18O) enriched substrates to inorganic phosphate (Pi) has been demonstrated for two enzyme-catalyzed reactions involved in folate biosynthesis and glutamylation. COOH-18O-labeled folate, methotrexate, and dihydropteroate, in addition to [17O]-glutamate, were synthesized
Qingguo Meng et al.
Environmental science & technology, 45(7), 3000-3005 (2011-03-03)
Due to the neurotoxicity of organophosphate (OP) pesticides and nerve agents synthesized as military or terror agents, their safe destruction and disposal is of considerable current importance. A representative OP, trimethyl phosphate (TMP), was adsorbed onto NaX zeolite, two mesoporous
K Sundararajan et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 56A(10), 1855-1867 (2000-09-16)
Trimethyl phosphate (TMP) and acetylene were codeposited in nitrogen and argon matrices and adducts of these species were identified using infrared spectroscopy. Formation of the adducts was evidenced by shifts in the vibrational frequencies of the modes involving the TMP
Johannes Bernarding et al.
Journal of the American Chemical Society, 128(3), 714-715 (2006-01-19)
In ultralow magnetic fields, liquid state nuclear magnetic resonance (NMR) spectra of homonuclear spin systems exhibit line widths dominated by their natural lifetime. Chemical shifts become negligible, and heteronuclear NMR spectra show predominantly the electron-mediated field-independent J-coupling. However, weak polarization

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service