Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

30-4260

Sigma-Aldrich

Triethylene glycol

SAJ first grade, ≥96.0%

Synonym(s):

Triglycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO(CH2CH2O)2CH2CH2OH
CAS Number:
Molecular Weight:
150.17
Beilstein/REAXYS Number:
969357
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
grade:
SAJ first grade
assay:
≥96.0%
bp:
125-127 °C/0.1 mmHg (lit.)
vapor pressure:
<0.01 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

SAJ first grade

vapor density

5.2 (vs air)

vapor pressure

<0.01 mmHg ( 20 °C)

assay

≥96.0%

form

liquid

autoignition temp.

699 °F

expl. lim.

9.2 %

availability

available only in Japan

dilution

(for analytical testing)

refractive index

n20/D 1.455 (lit.)

bp

125-127 °C/0.1 mmHg (lit.)

mp

−7 °C (lit.)

density

1.124 g/mL at 20 °C (lit.)

SMILES string

OCCOCCOCCO

InChI

1S/C6H14O4/c7-1-3-9-5-6-10-4-2-8/h7-8H,1-6H2

Inchi Key

ZIBGPFATKBEMQZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

345.2 °F

flash_point_c

174 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Babak Karimi et al.
Chemical communications (Cambridge, England), 47(27), 7686-7688 (2011-06-11)
A novel water-soluble NHC-Pd polymer with triethylene glycol legs was developed as a water soluble and highly recyclable catalyst that shows high catalytic activity in the Suzuki-Miyaura coupling of aryl chlorides in high yields in water at room temperature.
Amit Vaish et al.
Small (Weinheim an der Bergstrasse, Germany), 7(10), 1471-1479 (2011-05-04)
Soft-lithography-based techniques are widely used to fabricate microarrays. Here, the use of microcontact insertion printing is described, a soft-lithography method specifically developed for patterning at the dilute scales necessary for highly selective biorecognition. By carefully tuning the polar surface energy
Jianji Wang et al.
The journal of physical chemistry. B, 115(17), 4955-4962 (2011-04-13)
Material preparation in ionic liquids and environmental pollution control by ionic liquids are often closely dependent on the aggregation behavior of ionic liquids in solution. In the present work, conductivity, fluorescence probe, and dynamic light scattering techniques have been used
Rosa Doménech et al.
Biomacromolecules, 11(8), 2069-2078 (2010-08-10)
Assembly of the mature human immunodeficiency virus type 1 capsid involves the oligomerization of the capsid protein, CA. The C-terminal domain of CA, CTD, participates both in the formation of CA hexamers and in the joining of hexamers through homodimerization.
Barbara Sanavio et al.
ACS nano, 4(11), 6607-6616 (2010-10-21)
Nanopatterning of biomolecules on functionalized surfaces offers an excellent route for ultrasensitive protein immobilization, for interaction measurements, and for the fabrication of devices such as protein nanoarrays. An improved understanding of the physics and chemistry underlying the device properties and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service