30-3440
o-Toluidine
SAJ special grade, ≥99.0%
Synonym(s):
2-Aminotoluene, 2-Methylaniline
About This Item
grade
SAJ special grade
vapor density
3.7 (vs air)
vapor pressure
0.26 mmHg ( 25 °C)
assay
≥99.0%
autoignition temp.
899 °F
availability
available only in Japan
refractive index
n20/D 1.572 (lit.)
bp
199-200 °C (lit.)
89-90 °C/11 mmHg (lit.)
mp
−28 °C (lit.)
density
1.008 g/mL at 25 °C (lit.)
SMILES string
Cc1ccccc1N
InChI
1S/C7H9N/c1-6-4-2-3-5-7(6)8/h2-5H,8H2,1H3
InChI key
RNVCVTLRINQCPJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
signalword
Danger
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Carc. 1B - Eye Irrit. 2
Storage Class
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
wgk_germany
WGK 3
flash_point_f
185.0 °F - closed cup
flash_point_c
85 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service