Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

30-2850

Sigma-Aldrich

Titanium(IV) chloride

SAJ first grade, ≥97.0%

Synonym(s):

TTC, Titanium tetrachloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TiCl4
CAS Number:
Molecular Weight:
189.68
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
SAJ first grade
assay:
≥97.0%
bp:
135-136 °C (lit.)
vapor pressure:
50 mmHg ( 55 °C)
9.6 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

SAJ first grade

vapor pressure

50 mmHg ( 55 °C)
9.6 mmHg ( 20 °C)

assay

≥97.0%

form

liquid

reaction suitability

reagent type: catalyst
core: titanium

availability

available only in Japan

dilution

(for analytical testing)

bp

135-136 °C (lit.)

mp

−25 °C (lit.)

density

1.73 g/mL at 20 °C (lit.)

SMILES string

Cl[Ti](Cl)(Cl)Cl

InChI

1S/4ClH.Ti/h4*1H;/q;;;;+4/p-4

InChI key

XJDNKRIXUMDJCW-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

pictograms

Skull and crossbonesCorrosion

signalword

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

target_organs

Respiratory system

supp_hazards

Storage Class

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hossein Noyan et al.
PloS one, 10(6), e0130658-e0130658 (2015-06-23)
To understand the molecular pathways underlying the cardiac preconditioning effect of short-term caloric restriction (CR). Lifelong CR has been suggested to reduce the incidence of cardiovascular disease through a variety of mechanisms. However, prolonged adherence to a CR life-style is
Ravit Madar et al.
Brain, behavior, and immunity, 48, 301-312 (2015-05-30)
Neuroinflammation in the central nervous system is detrimental for learning and memory, as evident form epidemiological studies linking developmental defects and maternal exposure to harmful pathogens. Postnatal infections can also induce neuroinflammatory responses with long-term consequences. These inflammatory responses can
Sofia-Iris Bibli et al.
Cardiovascular research, 106(3), 432-442 (2015-04-15)
H2S is known to confer cardioprotection; however, the pathways mediating its effects in vivo remain incompletely understood. The purpose of the present study is to evaluate the contribution of cGMP-regulated pathways in the infarct-limiting effect of H2S in vivo. Anaesthetized
Charles Dylan Turner et al.
Organic letters, 14(18), 4970-4973 (2012-09-08)
The Ti(IV)-catalyzed Ugi condensation of α-amino acids with electron-rich aromatic aldehydes performs adequately even with sterically demanding α-amino carboxylate salts. The reaction occurs diastereoselectively, in some cases with virtually complete diastereoselectivity. A stereochemical rationale for the reaction is proposed.
Soji Shimizu et al.
Chemical communications (Cambridge, England), 49(16), 1621-1623 (2013-01-23)
Pyrrolopyrrole aza-BODIPY analogues were synthesized from diketopyrrolopyrrole and heteroaromatic amines in the presence of titanium tetrachloride. These novel compounds exhibit intense absorption in the visible region and strong emission with high fluorescence quantum yields greater than 0.8.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service