Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

30-2260

Sigma-Aldrich

Thymol

SAJ first grade, ≥98.0%

Synonym(s):

2-Isopropyl-5-methylphenol, 5-Methyl-2-(1-methylethyl)phenol, 5-Methyl-2-isopropylphenol, IPMP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-[(CH3)2CH]C6H3-5-(CH3)OH
CAS Number:
Molecular Weight:
150.22
Beilstein/REAXYS Number:
1907135
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
grade:
SAJ first grade
assay:
≥98.0%
bp:
232 °C (lit.)
vapor pressure:
1 mmHg ( 64 °C)
Pricing and availability is not currently available.

grade

SAJ first grade

vapor pressure

1 mmHg ( 64 °C)

assay

≥98.0%

form

solid

availability

available only in Japan

dilution

(for analytical testing)

bp

232 °C (lit.)

mp

48-51 °C (lit.)

density

0.965 g/mL at 25 °C (lit.)

SMILES string

CC(C)c1ccc(C)cc1O

InChI

1S/C10H14O/c1-7(2)9-5-4-8(3)6-10(9)11/h4-7,11H,1-3H3

Inchi Key

MGSRCZKZVOBKFT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

WGK 2

flash_point_f

219.2 °F

flash_point_c

104 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohammad Reza Youssefi et al.
Molecules (Basel, Switzerland), 24(11) (2019-06-04)
One of the most important causative agents of visceral leishmaniasis (VL) is Leishmania infantum, which is mainly spread by Phlebotomus and Lutzomyia sandflies in the Old and New World, respectively. Novel and effective drugs to manage this neglected vector-borne disease
C Benchaar
Journal of dairy science, 104(1), 324-336 (2020-11-03)
Phenolic compounds and essential oils with high content of phenolic compounds have been reported to exert antimicrobial activities in vitro. The objective of this study was to determine the effects of dairy cow diet supplementation with thyme oil and its
M Maggiore et al.
Parasitology international, 64(5), 435-440 (2015-06-23)
Cystic echinococcosis is a zoonotic disease caused by the larval stage of the cestode Echinococcus granulosus. The drugs commonly used against cystic echinococcosis are benzimidazoles. Unfortunately, 20%-40% of cases do not respond favorably to such chemotherapy. Consequently, the search of
Hiroko Ohwada et al.
Research in developmental disabilities, 34(1), 650-655 (2012-11-06)
As the life expectancy of people with intellectual disability (ID) increases, it is becoming necessary to understand factors affecting survival. However, predictors that are typically assessed among healthy people have not been examined. Predictors of all-cause mortality, including blood, urine
María C Elissondo et al.
Acta tropica, 125(3), 251-257 (2012-12-25)
Scolicidal solutions remain indispensable in the treatment of hydatid cyst disease. Properties of an ideal solution would be inexpensiveness and the promotion of a rapid and complete scolicidal effect with an absence of local and systemic side effects. From this

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service