Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

30-0635

Sigma-Aldrich

Tetrachloroethylene

SAJ first grade, ≥98.0%

Synonym(s):

PCE, Perchloroethylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CCl2=CCl2
CAS Number:
Molecular Weight:
165.83
Beilstein/REAXYS Number:
1361721
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
grade:
SAJ first grade
assay:
≥98.0%
bp:
121 °C (lit.)
vapor pressure:
13 mmHg ( 20 °C)
19 mmHg ( 25 °C)
Pricing and availability is not currently available.

grade

SAJ first grade

vapor density

5.83 (vs air)

vapor pressure

13 mmHg ( 20 °C)
19 mmHg ( 25 °C)

assay

≥98.0%

form

liquid

availability

available only in Japan

dilution

(for analytical testing)

refractive index

n20/D 1.505 (lit.)

bp

121 °C (lit.)

mp

−22 °C (lit.)

density

1.623 g/mL at 25 °C (lit.)

SMILES string

Cl\C(Cl)=C(\Cl)Cl

InChI

1S/C2Cl4/c3-1(4)2(5)6

InChI key

CYTYCFOTNPOANT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Warning

Hazard Classifications

Aquatic Chronic 2 - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

No data available

flash_point_c

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Larissa J Maier et al.
PloS one, 10(6), e0129805-e0129805 (2015-06-25)
The ongoing bioethical debate on pharmacological cognitive enhancement (PCE) in healthy individuals is often legitimated by the assumption that PCE will widely spread and become desirable for the general public in the near future. This assumption was questioned as PCE
Dusty R V Berggren et al.
Environmental science & technology, 47(4), 1879-1886 (2013-01-16)
Results are presented from a chemostat study where the reductive dehalogenation of PCE was evaluated in the absence and presence of sulfate. Two chemostats inoculated with the Point Mugu culture, which contains strains of Dehalococcoides mccartyi, were operated at a
Annika S Fjordbøge et al.
Journal of contaminant hydrology, 140-141, 56-66 (2012-09-27)
Field investigations on the effects of ZVI-Clay soil mixing were conducted at a small DNAPL source zone with PCE as the parent compound. In a one-year monitoring program, soil samples were collected at three horizontal sampling planes (2.5, 5.0 and
Charline Wiegert et al.
Environmental science & technology, 46(20), 10918-10925 (2012-09-20)
Chlorinated ethenes (CEs) are ubiquitous groundwater contaminants, yet there remains a need for a method to efficiently monitor their in situ degradation. We report here the first field application of combined stable carbon and chlorine isotope analysis of tetrachloroethene (PCE)
Biological actions and interactions of tetrachloroethylene.
D Reichert
Mutation research, 123(3), 411-429 (1983-12-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service