Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

28-5030

Sigma-Aldrich

Stearic acid

SAJ first grade, ≥90.0%

Synonym(s):

1-Heptadecanecarboxylic acid, C18:0, Cetylacetic acid, NSC 25956, NSC 261168, Octadecanoic acid, Stearophanic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)16COOH
CAS Number:
Molecular Weight:
284.48
Beilstein/REAXYS Number:
608585
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

grade

SAJ first grade

vapor pressure

1 mmHg ( 173.7 °C)

assay

≥90.0%

form

solid

availability

available only in Japan

bp

361 °C (lit.)

mp

67-72 °C (lit.)

density

0.845 g/cm3

functional group

carboxylic acid

storage temp.

room temp

SMILES string

CCCCCCCCCCCCCCCCCC(O)=O

InChI

1S/C18H36O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h2-17H2,1H3,(H,19,20)

InChI key

QIQXTHQIDYTFRH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

nwg

flash_point_f

392.0 °F - Cleveland open cup

flash_point_c

200 °C - Cleveland open cup

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Piret Arukuusk et al.
Biochimica et biophysica acta, 1828(5), 1365-1373 (2013-01-30)
Harnessing of a branched structure is a novel approach in the design of cell-penetrating peptides and it has provided highly efficient transfection reagents for intracellular delivery of nucleic acids. The new stearylated TP10 analogs, NickFects, condense plasmid DNA, splice correcting
Qian Wan et al.
Nanoscale, 5(2), 744-752 (2012-12-12)
As a versatile gene vector, minicircle DNA (mcDNA) has a great potential for gene therapy. However, some serious challenges remain, such as to effectively deliver mcDNA into targeted cells/tissues and to non-invasively monitor the delivery of the mcDNA. Superparamagnetic iron
T A Doran et al.
American journal of obstetrics and gynecology, 133(3), 302-307 (1979-02-01)
Lecithin/sphingomyelin (L/S) ratio, creatinine, percentage of lipid-positive cells, palmitic acid, palmitic acid/stearic acid (P/S) ratio, and total cortisol were analyzed as tests for fetal maturity and fetal pulmonary maturity in 164 samples of amniotic fluid from 121 patients. Fifty samples
S M Grundy
The American journal of clinical nutrition, 60(6 Suppl), 986S-990S (1994-12-01)
Stearic acid is a long-chain saturated fatty acid. However, in contrast with other saturated fatty acids, stearic acid apparently does not raise serum cholesterol concentrations. Studies carried out three decades ago provided strong suggestive evidence that this was the case.
A M Turpeinen et al.
Arteriosclerosis, thrombosis, and vascular biology, 18(2), 316-322 (1998-03-04)
The effects of stearic acid (C18:0) and trans-fatty acids (trans-FAs) on measures of platelet function and prostacyclin (PGI2) production are poorly understood in humans. In this controlled dietary study, platelet function and endothelial PGI2 production were studied in healthy humans

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service