Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

28-4960

Sigma-Aldrich

Starch from potato

CP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H10O5)n
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352300
Pricing and availability is not currently available.

biological source

potato

grade

CP

form

solid

availability

available only in Japan

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Proceedings of the 8th Starch Digestion Consortium Workshop, December 21-22, 2011, Baylor, Texas.
Journal of pediatric gastroenterology and nutrition, 55 Suppl 2, S1-47 (2013-04-27)
L V Obukhova et al.
Genetika, 49(3), 337-342 (2013-06-13)
The genetic control of grain hardness and its association with the specific friabilin content on starch granules of common wheat cultivars and lines with intervarietal substitutions of homeologous group-5 chromosomes were studied. A significant correlation was revealed between the technological
FDA delays decision to remove powdered gloves.
Richard F Edlich et al.
JAMA surgery, 148(5), 406-407 (2013-05-17)
Subramanian Saravanan et al.
The Journal of nutrition, 143(6), 781-787 (2013-04-26)
This study investigated the hypothesis that the voluntary feed intake in fish is regulated by diet-induced differences in oxygen use. Four diets were prepared with a similar digestible protein:digestible energy ratio (18 mg/kJ), but which differed in the composition of
Amanda Bortolini Silveira et al.
PLoS genetics, 9(4), e1003437-e1003437 (2013-04-18)
Epigenetic variation, such as heritable changes of DNA methylation, can affect gene expression and thus phenotypes, but examples of natural epimutations are few and little is known about their stability and frequency in nature. Here, we report that the gene

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service