Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

28-4780

Sigma-Aldrich

D-Sorbitol

SAJ first grade, ≥97.0%

Synonym(s):

D-Glucitol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14O6
CAS Number:
Molecular Weight:
182.17
Beilstein/REAXYS Number:
1721899
EC Number:
MDL number:
UNSPSC Code:
41116130
PubChem Substance ID:
Pricing and availability is not currently available.

grade

SAJ first grade

vapor density

<1 (vs air)

vapor pressure

<0.1 mmHg ( 25 °C)

assay

≥97.0%

availability

available only in Japan

mp

98-100 °C (lit.)

SMILES string

OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)CO

InChI

1S/C6H14O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3-12H,1-2H2/t3-,4+,5-,6-/m1/s1

InChI key

FBPFZTCFMRRESA-JGWLITMVSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Youngkook Kwon et al.
ChemSusChem, 6(3), 455-462 (2013-01-25)
This Full Paper addresses the electrocatalytic hydrogenation of glucose to sorbitol or 2-deoxysorbitol on solid metal electrodes in neutral media. Combining voltammetry and online product analysis with high-performance liquid chromatography (HPLC), provides both qualitative and quantitative information regarding the reaction
Anna Boccaccio et al.
Cellular and molecular life sciences : CMLS, 71(21), 4275-4283 (2014-04-29)
Two-pore channel proteins (TPC) encode intracellular ion channels in both animals and plants. In mammalian cells, the two isoforms (TPC1 and TPC2) localize to the endo-lysosomal compartment, whereas the plant TPC1 protein is targeted to the membrane surrounding the large
J M van Griensven et al.
Clinical pharmacology and therapeutics, 58(6), 631-640 (1995-12-01)
To examine the effect of diabetes mellitus on the pharmacokinetics of tolrestat and to investigate its effect on red blood cell sorbitol levels according to a new pharmacodynamic model for this class of drugs. Single and multiple doses of tolrestat
Beau Op de Beeck et al.
ChemSusChem, 6(1), 199-208 (2013-01-12)
The catalytic valorization of cellulose is currently subject of intense research. Isosorbide is among the most interesting products that can be formed from cellulose as it is a potential platform molecule and can be used for the synthesis of a
Reddy Ranjith K Sama et al.
Journal of cellular physiology, 228(11), 2222-2231 (2013-04-30)
FUsed in Sarcoma/Translocated in LipoSarcoma (FUS/TLS or FUS) has been linked to several biological processes involving DNA and RNA processing, and has been associated with multiple diseases, including myxoid liposarcoma and amyotrophic lateral sclerosis (ALS). ALS-associated mutations cause FUS to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service