Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

28-4590

Sigma-Aldrich

Hypo alcohol

in ethanol

Synonym(s):

Sodium thiosulfate solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2S2O3
CAS Number:
Molecular Weight:
158.11
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Pricing and availability is not currently available.

form

solution

availability

available only in Japan

concentration

in ethanol

SMILES string

[Na+].[Na+].[O-]S([O-])(=O)=S

InChI

1S/2Na.H2O3S2/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

AKHNMLFCWUSKQB-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemical peritonitis after intraperitoneal sodium thiosulfate.
D R Gupta et al.
Peritoneal dialysis international : journal of the International Society for Peritoneal Dialysis, 32(2), 220-222 (2012-03-03)
Melani Sooriyaarachchi et al.
Metallomics : integrated biometal science, 5(3), 197-207 (2013-02-28)
Cis-Platin (CP) is a remarkably effective Pt-based anticancer drug, but it also exhibits severe toxic side-effects, including nephrotoxicity and ototoxicity. Previous studies conducted with mammalian model organisms have clearly demonstrated that the intravenous administration of N-acetyl-L-cysteine (NAC) or sodium thiosulfate
V M Smith et al.
Clinical and experimental dermatology, 37(8), 874-878 (2012-05-03)
Calciphylaxis is a rare and potentially life-threatening condition. It is thought to result from arterial calcification causing complete vascular occlusion and subsequent cutaneous infarction. Most often, it is a complication of end-stage renal failure or hyperparathyroidism; without either of these
C Resgalla et al.
Chemosphere, 89(1), 102-107 (2012-05-26)
Since the development of the TIE (Toxicity Identification and Evaluation) in 1988 it has been assumed that the capacity of EDTA and sodium thiosulfate to complex some metals, and thus remove their toxicity, can be applied to both freshwater and
Melani Sooriyaarachchi et al.
Metallomics : integrated biometal science, 4(9), 960-967 (2012-07-31)
The anticancer drug cis-platin (CP) is widely used to treat patients, but it is also associated with significant side effects, including nephrotoxicity. Given that this metallodrug is intravenously (iv) administered, its biotransformations in the bloodstream are likely to be involved

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service