Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

28-4460

Sigma-Aldrich

Sodium tetraborate decahydrate

99.5-100.5%

Synonym(s):

Borax decahydrate, Sodium borate decahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2B4O7 · 10H2O
CAS Number:
Molecular Weight:
381.37
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
assay:
99.5-100.5%
grade:
SAJ special grade
form:
crystalline
Pricing and availability is not currently available.

grade

SAJ special grade

assay

99.5-100.5%

form

crystalline

availability

available only in Japan

pH

9.2 (10 g/L)

density

1.73 g/mL at 25 °C (lit.)

SMILES string

O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-]B1Ob2ob([O-])ob(O1)o2

InChI

1S/B4O7.2Na.10H2O/c5-1-7-3-9-2(6)10-4(8-1)11-3;;;;;;;;;;;;/h;;;10*1H2/q-2;2*+1;;;;;;;;;;

InChI key

CDMADVZSLOHIFP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Suitability

pH standard solution

Legal Information

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Repr. 1B

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eunice F S Vieira et al.
Biomacromolecules, 9(4), 1195-1199 (2008-03-19)
Oxidized alginate (ADA) and oxidized alginate blended with chitosan (ADA-Chit) were prepared in the presence of borax and CaCl 2, and their interactions with an antifolate drug, pyrimethamine (PYR), have been investigated. Tablets with a mean diameter of 1.2 +/-
Hasan Turkez
Journal of applied toxicology : JAT, 28(5), 658-664 (2007-11-21)
Titanium dioxide (TiO(2)) is a potential carcinogenic/mutagenic agent although it is used in many areas including medical industries and cosmetics. Boron (as boric acid and borax) has also well-described biological effects and therapeutic benefits. In a previous study, sister-chromatid exchanges
Jian-Feng He et al.
Journal of chromatography. A, 1218(24), 3816-3821 (2011-05-14)
The paper presents an on-line transient moving chemical reaction boundary (MCRB) method for simply but efficiently stacking analytes in capillary electrophoresis (CE). The CE technique was developed for a rapid determination of fumaric and maleic acid. Based on the theory
Xuan Wei et al.
Journal of chromatography. A, 1281, 148-154 (2013-02-13)
With an increasing concern on food safety, fast screening of residues of widespread herbicides becomes necessary. Herein we report a microchip electrophoresis system with laser induced fluorescence (LIF) detection for rapid and sensitive analysis of glyphosate (GLYP) and glufosinate (GLUF)
Marco Frasconi et al.
Langmuir : the ACS journal of surfaces and colloids, 25(18), 11097-11104 (2009-08-22)
A highly stable biological film was prepared by casting an aqueous dispersion of protein and composite hydrogel obtained from the polysaccharide Scleroglucan (Sclg) and borax as a cross-linking agent. Heme proteins, such as hemoglobin (Hb), myoglobin (Mb), and horseradish peroxidase

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service