Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

28-4130

Sigma-Aldrich

Sodium silicate hydrate

CP

Synonym(s):

Sodium silicate solution, Sodium trisilicate solution, Water glass

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2O(SiO2)x · xH2O
EC Number:
UNSPSC Code:
12352300
grade:
CP
form:
solid
Pricing and availability is not currently available.

grade

CP

form

solid

availability

available only in Japan

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service