Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

28-3350

Sigma-Aldrich

Phosphate glass, water insoluble

SAJ first grade, 65-70% P2O5 basis

Synonym(s):

Insoluble sodium metaphosphate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352300
grade:
SAJ first grade
form:
solid
Pricing and availability is not currently available.

grade

SAJ first grade

form

solid

availability

available only in Japan

concentration

65-70% P2O5

pH

5.7 (20 °C, 10 g/L)

storage temp.

15-25°C

InChI

1S/Na.HO3P/c;1-4(2)3/h;(H,1,2,3)/q+1;/p-1

InChI key

AQMNWCRSESPIJM-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service