Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

28-2920

Sigma-Aldrich

Sodium hydrosulfide hydrate

CP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaSH·xH2O
CAS Number:
Molecular Weight:
56.06 (anhydrous basis)
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Pricing and availability is not currently available.

grade

CP

form

solid

availability

available only in Japan

mp

52-54 °C (lit.)

storage temp.

15-25°C

SMILES string

[Na]S.[H]O[H]

InChI

1S/Na.H2O.H2S/h;2*1H2/q+1;;/p-1

InChI key

ZNKXTIAQRUWLRL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Skin Corr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 2

flash_point_f

194.0 °F - closed cup

flash_point_c

90 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masaki Nasu et al.
Journal of thoracic oncology : official publication of the International Association for the Study of Lung Cancer, 10(4), 565-576 (2015-02-07)
Breast cancer 1-associated protein 1 (BAP1) is a nuclear deubiquitinase that regulates gene expression, transcription, DNA repair, and more. Several findings underscore the apparent driver role of BAP1 in malignant mesothelioma (MM). However, the reported frequency of somatic BAP1 mutations
Sourav Kundu et al.
Nitric oxide : biology and chemistry, 46, 172-185 (2015-02-11)
Previously we reported that matrix metalloproteinase-9 (MMP-9) plays an important role in extracellular matrix (ECM) remodeling in diabetic kidney. Induction of NMDA-R and dysregulation of connexins (Cxs) were also observed. We concluded that this was due to decreased H2S production
Takashi Sonobe et al.
Clinical toxicology (Philadelphia, Pa.), 53(6), 525-539 (2015-05-13)
Hydrogen sulfide (H2S) intoxication produces an acute depression in cardiac contractility-induced circulatory failure, which has been shown to be one of the major contributors to the lethality of H2S intoxication or to the neurological sequelae in surviving animals. Methylene blue
Sang Jun Han et al.
Nephrology, dialysis, transplantation : official publication of the European Dialysis and Transplant Association - European Renal Association, 30(9), 1497-1506 (2015-07-05)
Progression of acute kidney injury to chronic kidney disease (CKD) is associated with inadequate recovery of damaged kidney. Hydrogen sulfide (H2S) regulates a variety of cellular signals involved in cell death, differentiation and proliferation. This study aimed to identify the
Philippe Haouzi et al.
Toxicological sciences : an official journal of the Society of Toxicology, 141(2), 493-504 (2014-07-13)
Hydrogen sulphide (H2S), a chemical hazard in oil and gas production, has recently become a dreadful method of suicide, posing specific risks and challenges for the first responders. Currently, there is no proven effective treatment against H2S poisoning and its

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service