Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

28-2840

Sigma-Aldrich

Disodium beta-glycerophosphate pentahydrate

SAJ special grade, ≥98.0%

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352300
grade:
SAJ special grade
assay:
≥98.0%
Pricing and availability is not currently available.

grade

SAJ special grade

assay

≥98.0%

form

solid

availability

available only in Japan

dilution

(for analytical testing)

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service