Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

28-2220

Sigma-Aldrich

Sodium carbonate decahydrate

SAJ first grade, ≥99.0%

Synonym(s):

Sal soda, Soda

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2CO3 · 10H2O
CAS Number:
Molecular Weight:
286.14
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
assay:
≥99.0%
grade:
SAJ first grade
form:
solid

grade

SAJ first grade

assay

≥99.0%

form

solid

availability

available only in Japan

pH

11.0-12.0 (25 °C, 50 g/L)

storage temp.

15-25°C

SMILES string

O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-]C([O-])=O

InChI

1S/CH2O3.2Na.10H2O/c2-1(3)4;;;;;;;;;;;;/h(H2,2,3,4);;;10*1H2/q;2*+1;;;;;;;;;;/p-2

InChI key

XYQRXRFVKUPBQN-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hagen Frickmann et al.
PloS one, 10(8), e0135923-e0135923 (2015-08-14)
The quality of microbiological diagnostic procedures depends on pre-analytic conditions. We compared the results of 16S rRNA gene PCR and sequencing from automatically incubated blood culture materials from tropical Ghana with the results of cultural growth after automated incubation. Real-time
Nadine Schlueter et al.
Archives of oral biology, 57(9), 1176-1182 (2012-05-05)
The eroded organic dentine matrix is remarkably resistant to mechanical impacts. Additional brushing abrasion of eroded dentine has only limited influence on tissue loss. Digestive enzymes (e.g., pepsin, trypsin) that can reach the oral cavity during reflux or vomiting can
Tingting Wu et al.
Environmental science & technology, 46(4), 2291-2298 (2012-02-01)
Many advanced oxidation processes involve addition of hydrogen peroxide (H(2)O(2)) with the aim of generating hydroxyl radicals to oxidize organic contaminants in water. However, chemical oxygen demand, a common measure of gross residual organic contamination, is subject to interference from
Ning Bu et al.
Ecotoxicology and environmental safety, 78, 35-40 (2011-12-06)
Endophyte infected and uninfected seedlings of rice (Oryza sativa L.) were subjected to five different levels of Na(2)CO(3) for 2 weeks. Under both Na(2)CO(3) stress as well as no stress, endophyte-infected plants were higher for above-ground dry weight and shoot
Sandra Nilsson et al.
PloS one, 7(11), e46980-e46980 (2012-11-28)
During the intraerythrocytic development of Plasmodium falciparum, the malaria parasite remodels the host cell cytosol by inducing membranous structures termed Maurer's clefts and inserting parasite proteins into the red blood cell cytoskeleton and plasma membrane. Pf332 is the largest known

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service