Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

28-2010

Sigma-Aldrich

Sodium tetraborate

SAJ first grade, ≥99.0%

Synonym(s):

Borax, fused

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2B4O7
CAS Number:
Molecular Weight:
201.22
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

SAJ first grade

assay

≥99.0%

form

solid

availability

available only in Japan

mp

741 °C (lit.)

density

2.367 g/mL at 25 °C (lit.)

storage temp.

15-25°C

SMILES string

[Na+].[Na+].[O-]B1Ob2ob([O-])ob(O1)o2

InChI

1S/B4O7.2Na/c5-1-7-3-9-2(6)10-4(8-1)11-3;;/q-2;2*+1

InChI key

UQGFMSUEHSUPRD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Repr. 1B

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dušan Koval et al.
Analytical biochemistry, 413(1), 8-15 (2011-02-09)
Two capillary electrophoretic methods were developed and evaluated for measurement of glycated hemoglobin A1c (HbA1c). First, a capillary electrophoresis analysis is performed with a sodium tetraborate buffer (pH 9.3) as background electrolyte in a neutrally coated capillary. HbA1c is separated
Shridhar K Sathe et al.
Journal of agricultural and food chemistry, 57(17), 7846-7856 (2009-08-07)
The solubility of almond, Brazil nut, cashew nut, hazelnut, macadamia, pecan, pine nut, pistachio, walnut, and peanut proteins in several aqueous solvents was qualitatively and quantitatively assessed. In addition, the effects of extraction time and ionic strength on protein solubility
Qianqian Zhu et al.
Journal of chromatography. A, 1246, 35-39 (2012-03-03)
A technique known as field enhancement sample stacking (FESS) and capillary electrophoresis (CE) separation has been developed to analyze and detect organic acids in the three traditional Chinese medicines (such as Portulaca oleracea L., Crataegus pinnatifida and Aloe vera L.).
Shao-Qiang Hu et al.
Electrophoresis, 34(2), 260-268 (2012-11-20)
Based on the investigation of the effect of microemulsion charge on the chiral separation, a new chiral separation method with MEEKC employing neutral microemulsion was established. The method used a microemulsion containing 3.0% (w/v) neutral surfactant Tween 20 and 0.8%
Jamshed Iqbal et al.
Journal of chromatography. A, 1218(29), 4764-4771 (2011-06-15)
A simple, efficient, and highly sensitive in-line CE method was developed for the characterization and for inhibition studies of the nucleoside-metabolizing enzymes purine nucleoside phosphorylase (PNP) and adenosine deaminase (ADA) present in membrane preparations of human 1539 melanoma cells. After

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service