Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

28-1789

Sigma-Aldrich

Sodium azide

JIS special grade, ≥98.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaN3
CAS Number:
Molecular Weight:
65.01
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
assay:
≥98.0%
grade:
JIS special grade
form:
solid
Pricing and availability is not currently available.

grade

JIS special grade

vapor pressure

0.01 hPa ( 20 °C)

assay

≥98.0%

form

solid

availability

available only in Japan

pH

10 (25 °C, 65 g/L)

storage temp.

15-25°C

SMILES string

[Na]N=[N+]=[N-]

InChI

1S/N3.Na/c1-3-2;/q-1;+1

InChI key

PXIPVTKHYLBLMZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2 Oral

target_organs

Brain

supp_hazards

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gilles Lambert et al.
Journal of the American College of Cardiology, 63(22), 2365-2373 (2014-03-19)
Do elevated proprotein convertase subtilisin/kexin type 9 (PCSK9) levels constitute an even greater risk for patients who already have reduced low-density lipoprotein receptor (LDLR) levels, such as those with heterozygous familial hypercholesterolemia (HeFH)? As a circulating inhibitor of LDLR, PCSK9
Liyi Huang et al.
Lasers in surgery and medicine, 44(6), 490-499 (2012-07-05)
Antimicrobial photodynamic therapy (APDT) employs a non-toxic photosensitizer (PS) and visible light, which in the presence of oxygen produce reactive oxygen species (ROS), such as singlet oxygen ((1) O(2), produced via Type II mechanism) and hydroxyl radical (HO(.), produced via
Andrew J Kassianos et al.
Kidney international, 87(6), 1153-1163 (2015-01-15)
Chemokines play pivotal roles in tissue recruitment and retention of leukocytes, with CX3CR1 recently identified as a chemokine receptor that selectively targets mouse kidney dendritic cells (DCs). We have previously demonstrated increased tubulointerstitial recruitment of human transforming growth factor-β (TGF-β)-producing
Luisa M Villar et al.
Annals of neurology, 76(2), 231-240 (2014-06-10)
To identify a biomarker distinguishing patients who, despite a primary progressive multiple sclerosis (PPMS) clinical course, may nonetheless benefit from immune therapy. The presence or absence of both immunoglobulin (Ig) G and IgM oligoclonal bands (OCB) was blindly examined in
Andre R Massensini et al.
Acta biomaterialia, 27, 116-130 (2015-09-01)
Biomaterials composed of mammalian extracellular matrix (ECM) promote constructive tissue remodeling with minimal scar tissue formation in many anatomical sites. However, the optimal shape and form of ECM scaffold for each clinical application can vary markedly. ECM hydrogels have been

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service