Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

28-0990

Sigma-Aldrich

Silicon dioxide

SAJ first grade

Synonym(s):

Cristobalite, Quartz, Sand, Silica

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SiO2
CAS Number:
Molecular Weight:
60.08
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
Pricing and availability is not currently available.

grade

SAJ first grade

form

solid

availability

available only in Japan

refractive index

n20/D 1.544 (lit.)

mp

1610 °C (lit.)

density

2.6 g/mL at 25 °C (lit.)

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yijun Yao et al.
Journal of hazardous materials, 298, 221-231 (2015-06-10)
In 2002, U.S. EPA proposed a general buffer zone of approximately 100 feet (30 m) laterally to determine which buildings to include in vapor intrusion (VI) investigations. However, this screening distance can be threatened by factors such as extensive surface
Keren B Turton et al.
PloS one, 10(5), e0127243-e0127243 (2015-05-20)
Signal transducer and activator of transcription 3 (STAT3) is a key mediator of leukocyte differentiation and proliferation. The 3' end of STAT3 transcripts is subject to two alternative splicing events. One results in either full-length STAT3α or in STAT3β, which
C Uhrenfeldt et al.
Optics express, 23(11), A525-A538 (2015-06-16)
Plasmonic resonances in metal nanoparticles are considered candidates for improved thin film Si photovoltaics. In periodic arrays the influence of collective modes can enhance the resonant properties of such arrays. We have investigated the use of periodic arrays of Al
Marcelo Ramalho-Ortigão et al.
Parasites & vectors, 8, 298-298 (2015-05-30)
The Phlebotomus papatasi salivary protein PpSP15 was shown to protect mice against Leishmania major, suggesting that incorporation of salivary molecules in multi-component vaccines may be a viable strategy for anti-Leishmania vaccines. Here, we investigated PpSP15 predicted amino acid sequence variability
Soania Mathur et al.
Journal of Parkinson's disease, 5(2), 263-268 (2015-02-28)
Despite an urgent need for new medications, clinical trials in Parkinson's have a relatively low rate of success. Although many reasons have been proposed for this, the opinions of patients and scientists, the two principal stakeholders, have not been widely

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service