Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

26-0190

Sigma-Aldrich

Quinoline

JIS special grade, ≥95.0%

Synonym(s):

1-Benzazine, 2,3-Benzopyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7N
CAS Number:
Molecular Weight:
129.16
Beilstein/REAXYS Number:
107477
MDL number:
UNSPSC Code:
12352210
PubChem Substance ID:
grade:
JIS special grade
assay:
≥95.0%
bp:
113-114 °C/11 mmHg (lit.)
237 °C (lit.)
vapor pressure:
0.07 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

JIS special grade

vapor density

4.5 (vs air)

vapor pressure

0.07 mmHg ( 20 °C)

assay

≥95.0%

form

liquid

autoignition temp.

896 °F

availability

available only in Japan

dilution

(for analytical testing)

refractive index

n20/D 1.625 (lit.)

bp

113-114 °C/11 mmHg (lit.)
237 °C (lit.)

mp

−17-−13 °C (lit.)

density

1.093 g/mL at 25 °C (lit.)

SMILES string

c1ccc2ncccc2c1

InChI

1S/C9H7N/c1-2-6-9-8(4-1)5-3-7-10-9/h1-7H

InChI key

SMWDFEZZVXVKRB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Chronic 2 - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 2

flash_point_f

221.0 °F

flash_point_c

105 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jin Hoon Kim et al.
Dalton transactions (Cambridge, England : 2003), 42(15), 5500-5507 (2013-02-22)
Zinc sensors based on 2,3-dipicolylamine (DPA) and quinoline have been synthesized. They fluoresced in the presence of Zn(2+) and remained fluorescent when other metal ions were present. Fluorescence enhancement of the sensors was not seen for most other metal ions.
Narendra Reddy Chereddy et al.
Dalton transactions (Cambridge, England : 2003), 41(38), 11753-11759 (2012-08-21)
Two new rhodamine based probes 1 and 2 for the detection of Fe(3+) were synthesized and their selectivity towards Fe(3+) ions in the presence of other competitive metal ions tested. The probe 1 formed a coloured complex with Fe(3+) as
Fang-Fang Wang et al.
Organic & biomolecular chemistry, 10(43), 8605-8608 (2012-10-09)
A practical Brønsted acid promoted benzylic C-H functionalization of 2-methylazaarenes and nucleophilic addition to aldehydes was developed in good to excellent yields. A six-membered hydrogen-bonding transition state is proposed to be crucial for the reaction. Ready availability of the two
Yu Zhang et al.
Dalton transactions (Cambridge, England : 2003), 41(38), 11776-11782 (2012-08-21)
A series of carboxamidoquinoline-based fluorescent sensors (the AQZ family) were synthesized and characterized. The AQZ family members were highly soluble in water and showed good selectivity for Zn(2+)via enhanced fluorescence in aqueous buffer solution. Fluorescence signals could be tuned from
Margarita A Gonzalez et al.
Inorganic chemistry, 51(21), 11930-11940 (2012-10-24)
The unusual role of CO as a signaling molecule in several physiological pathways has spurred research in the area of synthesizing new CO-releasing molecules (CORMs) as exogenous CO donors. Auxiliary control on CO delivery can be achieved if CO can

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service