Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

24-5080

Sigma-Aldrich

Potassium oxalate monohydrate

SAJ first grade, ≥99.0%

Synonym(s):

Ethanedioic acid, Oxalic acid potassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(COOK)2 · H2O
CAS Number:
Molecular Weight:
184.23
Beilstein/REAXYS Number:
3752576
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
assay:
≥99.0%
grade:
SAJ first grade
form:
powder
Pricing and availability is not currently available.

grade

SAJ first grade

assay

≥99.0%

form

powder

availability

available only in Japan

pH

7.0-8.5 (25 °C, 50 g/L)

SMILES string

O.[K+].[K+].[O-]C(=O)C([O-])=O

InChI

1S/C2H2O4.2K.H2O/c3-1(4)2(5)6;;;/h(H,3,4)(H,5,6);;;1H2/q;2*+1;/p-2

InChI key

QCPTVXCMROGZOL-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Sosvorová et al.
Physiological research, 63(4), 521-527 (2014-06-09)
Increased homocysteine levels in serum are typical features of neurodegenerative brain diseases including hydrocephalus. The most frequent therapeutic approach consists of the insertion of a shunt, connecting the brain ventricles to an alternative drainage site. To decide whether the patient

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service