Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

24-4890

Sigma-Aldrich

Potassium iodide

suitable for oxidant determination, ≥99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KI
CAS Number:
Molecular Weight:
166.00
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
assay:
≥99.5%
grade:
SAJ special grade
form:
solid
Pricing and availability is not currently available.

grade

SAJ special grade

vapor pressure

1 mmHg ( 745 °C)

assay

≥99.5%

form

solid

availability

available only in Japan

pH

~6.9 (20 °C, 50 g/L)

mp

681 °C (lit.)

suitability

suitable for oxidant determination

SMILES string

[K+].[I-]

InChI

1S/HI.K/h1H;/q;+1/p-1

InChI key

NLKNQRATVPKPDG-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

STOT RE 1 Oral

target_organs

Thyroid

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pietro Santulli et al.
Human reproduction (Oxford, England), 30(1), 49-60 (2014-11-08)
Are protein oxidative stress markers [thiols, advanced oxidation protein products (AOPP), protein carbonyls and nitrates/nitrites] in perioperative peritoneal fluid higher in women with histologically proven endometriosis when compared with endometriosis-free controls? Protein oxidative stress markers are significantly increased in peritoneal
Zhi-Qi Lao et al.
Organic & biomolecular chemistry, 10(39), 7869-7871 (2012-09-06)
We have developed a new KI-catalyzed method for the imidation of an sp(3) C-H bond adjacent to an amide nitrogen atom by using TBHP (tert-butyl hydroperoxide, 70% aqueous solution) as the oxidant. This novel procedure tolerated air and moisture and
S C Clement et al.
Pediatric blood & cancer, 60(11), 1833-1838 (2013-07-09)
Thyroid dysfunction has been reported in up to 52% of patients 1.4 years after treatment with (131) I-Metaiodobenzylguanidine (MIBG) in children with neuroblastoma (NBL), despite the use of potassium-iodide (KI). Our aim was to investigate if the incidence and severity
J B Sterling et al.
Journal of the American Academy of Dermatology, 43(4), 691-697 (2000-09-27)
Potassium iodide (KI) is a useful drug in the dermatologic armamentarium. It is successfully used for inflammatory dermatoses, most notably erythema nodosum, subacute nodular migratory panniculitis, nodular vasculitis, erythema multiforme, and Sweet's syndrome. KI is also successfully used for cutaneous
Kirsti Parikka et al.
Biomacromolecules, 13(8), 2418-2428 (2012-06-26)
Carboxylated, anionic polysaccharides were selectively prepared using a combination of enzymatic and chemical reactions. The galactose-containing polysaccharides studied were spruce galactoglucomannan, guar galactomannan, and tamarind galactoxyloglucan. The galactosyl units of the polysaccharides were first oxidized with galactose oxidase (EC 1.1.3.9)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service