Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

24-4530

Sigma-Aldrich

Potassium hexacyanoferrate(III)

JIS special grade, ≥99.0%

Synonym(s):

Potassium ferricyanide, Red prussiate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K3Fe(CN)6
CAS Number:
Molecular Weight:
329.24
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Pricing and availability is not currently available.

grade

JIS special grade

assay

≥99.0%

form

crystalline

reaction suitability

reagent type: catalyst
core: iron

availability

available only in Japan

pH

6.0-9.0 (25 °C, 329 g/L)

SMILES string

[K+].[K+].[K+].N#C[Fe-3](C#N)(C#N)(C#N)(C#N)C#N

InChI

1S/6CN.Fe.3K/c6*1-2;;;;/q;;;;;;-3;3*+1

InChI key

MIMJFNVDBPUTPB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation markEnvironment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2

supp_hazards

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liju Yang et al.
Biomedical microdevices, 13(2), 279-289 (2010-11-26)
Microlithographically fabricated interdigitated microsensor electrodes (IMEs) were cleaned, surface activated, chemically functionalized (amine) and derivatized with an Acrloyl-PEG-NHS to receive a spun-applied monomer cocktail of UV polymerizable monomer. IMEs were 2050.5, 1550.5, 1050.5 and 0550.5 possessing lines and spaces that
Daehee Kim et al.
ChemSusChem, 5(6), 1086-1091 (2012-05-10)
To scale-up microbial fuel cells (MFCs), installing multiple unit cells in a common reactor has been proposed; however, there has been a serious potential drop when connecting unit cells in series. To determine the source of the loss, a basic
Suqin Han et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 878(28), 2901-2904 (2010-09-08)
A novel capillary electrophoresis (CE) with chemiluminescence (CL) detection method for the determination of mitoxantrone (MTX) has been developed, which based on the CL reaction of potassium ferricyanide with luminol in sodium hydroxide medium sensitized by MTX. Under optimum analytical
Hideo Takakusa et al.
Drug metabolism and disposition: the biological fate of chemicals, 39(6), 1022-1030 (2011-03-03)
Lapatinib, an oral breast cancer drug, has recently been reported to be a mechanism-based inactivator of cytochrome P450 (P450) 3A4 and also an idiosyncratic hepatotoxicant. It was suggested that formation of a reactive quinoneimine metabolite was involved in mechanism-based inactivation
Lu Han et al.
Journal of pharmaceutical and biomedical analysis, 58, 141-145 (2011-10-15)
In this paper, a novel chemiluminescence (CL) system, 2-phenyl-4, 5-di (2-furyl) imidazole (PDFI)-potassium ferricyanide, for the determination of terbutaline sulfate was described. The method was based on enhancement of CL emission of PDFI-potassium ferricyanide system in the presence of terbutaline

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service