Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

24-4130

Sigma-Aldrich

Potassium L-tartrate monobasic

SAJ special grade, ≥99.5%

Synonym(s):

L(+)-Tartaric acid monopotassium salt, Potassium bitartrate, Potassium hydrogen L-tartrate, Potassium hydrogen tartrate, Tartaric acid monopotassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KO2CCH(OH)CH(OH)CO2H
CAS Number:
Molecular Weight:
188.18
Beilstein/REAXYS Number:
6119985
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
assay:
≥99.5%
grade:
SAJ special grade
form:
powder

grade

SAJ special grade

assay

≥99.5%

form

powder

availability

available only in Japan

pH

3.4-3.7 (20 °C)

density

1.954 g/mL at 25 °C (lit.)

SMILES string

[K+].O[C@H]([C@@H](O)C([O-])=O)C(O)=O

InChI

1S/C4H6O6.K/c5-1(3(7)8)2(6)4(9)10;/h1-2,5-6H,(H,7,8)(H,9,10);/q;+1/p-1/t1-,2-;/m1./s1

InChI key

KYKNRZGSIGMXFH-ZVGUSBNCSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

410.0 °F

flash_point_c

210 °C

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service