Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

24-3730

Sigma-Aldrich

Poly(propylene glycol)

CP, average Mw 400

Synonym(s):

PPG, Poly(propylene oxide)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H[OCH(CH3)CH2]nOH
CAS Number:
MDL number:
UNSPSC Code:
12162002
grade:
CP
vapor pressure:
<0.01 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

CP

vapor density

>1 (vs air)

vapor pressure

<0.01 mmHg ( 20 °C)

mol wt

average Mw 400

contains

130-190 ppm proprietary phenolic antioxidant

availability

available only in Japan

dilution

(for analytical testing)

SMILES string

CC(O)CO

InChI

1S/C6H14O3/c1-5(8)4-9-6(2)3-7/h5-8H,3-4H2,1-2H3

InChI key

DUFKCOQISQKSAV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Celeste R Brennecka et al.
Journal of biomedical materials research. Part B, Applied biomaterials, 100(5), 1298-1309 (2012-04-20)
This study examines the in vitro characteristics of a crosslinking polymer system for cerebral aneurysm embolization. The polymeric material is composed of poly(propylene glycol)diacrylate (PPODA) and pentaerythritol tetrakis(3-mercaptopropionate) (QT), formulated with the liquid contrast agents Conray™ or Omnipaque™ 300. The
Claudio Poggio et al.
Journal of endodontics, 38(9), 1239-1243 (2012-08-16)
The objective of this study was to evaluate and compare in vitro the decalcifying capability on root canal dentin of 4 different irrigating solutions. Twenty-five freshly extracted maxillary central incisors were selected. The canals were prepared to obtain a total
Zhiyong Zhao et al.
Chemical communications (Cambridge, England), 48(78), 9753-9755 (2012-08-25)
The designed DNA sequences can make DNA-b-PPO undergo in situ transition between diblock and triblock upon pH changes, consequently, induce a morphology-shifting from spherical micelles to nanofibers. This process is reversible and the assembled structures have been characterized by CD
Jessalyn Cortese et al.
Journal of the American Chemical Society, 134(8), 3671-3674 (2012-02-11)
We show here that complementary interactions can suppress mesoscopic order and thus lead to a counterintuitive change in material properties. We present results for telechelic supramolecular polymers based on poly(propylene oxide) (PPO), thymine (Thy), and diaminotriazine (DAT). The self-complementary systems
Omnidirectional printing of 3D microvascular networks.
Willie Wu et al.
Advanced materials (Deerfield Beach, Fla.), 23(24), H178-H183 (2011-03-26)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service