Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

24-2910

Sigma-Aldrich

Phthalic anhydride

SAJ first grade, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H4O3
CAS Number:
Molecular Weight:
148.12
Beilstein/REAXYS Number:
118515
EC Number:
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:
assay:
≥99.0%
grade:
SAJ first grade
form:
crystalline
Pricing and availability is not currently available.

grade

SAJ first grade

vapor density

5.1 (vs air)

vapor pressure

<0.01 mmHg ( 20 °C)

assay

≥99.0%

form

crystalline

autoignition temp.

1058 °F

expl. lim.

10.4 %

availability

available only in Japan

pH

2 (20 °C, 6 g/L)

bp

284 °C (lit.)

mp

131-134 °C (lit.)

SMILES string

O=C1OC(=O)c2ccccc12

InChI

1S/C8H4O3/c9-7-5-3-1-2-4-6(5)8(10)11-7/h1-4H

InChI key

LGRFSURHDFAFJT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

305.6 °F - DIN 51758

flash_point_c

152 °C - DIN 51758

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Micó-Tormos et al.
Journal of chromatography. A, 1203(1), 47-53 (2008-07-25)
The esterification of fatty alcohol ethoxylates (FAEs) with phthalic anhydride in 1,4-dioxane was studied. At 110 degrees C and in the presence of urea, which increased the reaction rate, esterification was completed in 60 min. The reaction yield did not
Carlton L Campbell et al.
The Journal of organic chemistry, 74(16), 6368-6370 (2009-07-11)
An efficient enantioselective synthesis of benzyl (1S,2R,4R)-4-(tert-butoxycarbonylamino)-2-(hydroxymethyl)cyclohexylcarbamate 2, an essential intermediate for a series of potent CCR2 antagonists, is described. The key step in the sequence is an iodolactamization to yield the highly functionalized (1R,2S,4S,5S)-tert-butyl 2-(benzyloxycarbonylamino)-4-iodo-7-oxo-6-azabicyclo[3.2.1]octane-6-carboxylate 11. An examination of
Sarah Quartier et al.
Contact dermatitis, 55(5), 257-267 (2006-10-10)
Copolymers or heteropolymers are large molecules with high molecular weights (>1000 D). They have been underestimated for a long time as to their sensitizing capacities. Allergic contact dermatitis to 6 copolymers in cosmetics and 1 in a medical dressing has
Yuichi Kajita et al.
Journal of the American Chemical Society, 130(51), 17226-17227 (2008-12-04)
An intermolecular nickel-catalyzed addition reaction has been developed where phthalic anhydrides react with alkynes to afford substituted isocoumarins. A mechanistic rationale is proposed, implying reductive elimination of Ni(0) promoted by ZnCl(2) cocatalyst as the key step of the catalytic cycle.
Yaoting Yu et al.
Artificial cells, blood substitutes, and immobilization biotechnology, 36(1), 63-73 (2008-02-23)
Agar-gel resin with tryptophan as ligand was synthesized as an adsorbent for the removal of Immunoglobulin E (IgE) in bronchial asthma. Factors affecting the adsorption properties of the adsorbent, such as the different kind of amino acid linked to the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service