Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

24-2890

Sigma-Aldrich

Phthalic acid

SAJ first grade, ≥99.0%

Synonym(s):

1,2-Benzenedicarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,2-(CO2H)2
CAS Number:
Molecular Weight:
166.13
Beilstein/REAXYS Number:
608199
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
Pricing and availability is not currently available.

grade

SAJ first grade

assay

≥99.0%

form

crystalline

availability

available only in Japan

mp

210-211 °C (dec.) (lit.)

SMILES string

OC(C1=C(C(O)=O)C=CC=C1)=O

InChI

1S/C8H6O4/c9-7(10)5-3-1-2-4-6(5)8(11)12/h1-4H,(H,9,10)(H,11,12)

InChI key

XNGIFLGASWRNHJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

334.4 °F

flash_point_c

168 °C

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christopher H Hurst et al.
Toxicological sciences : an official journal of the Society of Toxicology, 74(2), 297-308 (2003-06-14)
Phthalate esters are widely used as plasticizers in the manufacture of products made of polyvinyl chloride. Mono-(2-ethylhexyl)-phthalate (MEHP) induces rodent hepatocarcinogenesis by a mechanism that involves activation of the nuclear transcription factor peroxisome proliferator-activated receptor-alpha (PPARalpha). MEHP also activates PPAR-gamma
Paula J Lapinskas et al.
Toxicology, 207(1), 149-163 (2004-12-14)
Phthalate esters belong to a large class of compounds known as peroxisome proliferators (PP). PP include chemicals that activate different subtypes of the peroxisome proliferator-activated receptor (PPAR) family. The ability of phthalate esters and their metabolites to activate responses through
Russ Hauser
Seminars in reproductive medicine, 24(3), 156-167 (2006-06-29)
Scientific and public concern about the potential risk of environmental chemicals to male reproductive health has been heightened by reports of downward trends in semen quality, as well as increased rates of developmental urogenital tract anomalies and testicular cancer. Of
Martha M Téllez-Rojo et al.
The Science of the total environment, 461-462, 386-390 (2013-06-12)
Previous studies suggest that prenatal phthalate exposure affects neurodevelopment and behavior during the first years of life. To evaluate the effect of maternal urinary concentrations of phthalate metabolites during pregnancy on mental and psychomotor development in children 24-36 months of
Randi J Bertelsen et al.
Environmental health perspectives, 121(2), 251-256 (2012-11-21)
High-molecular-weight phthalates in indoor dust have been associated with asthma in children, but few studies have evaluated phthalate biomarkers in association with respiratory outcomes. We explored the association between urinary concentrations of phthalate metabolites and current asthma. In a cross-sectional

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service