Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

24-1560

Sigma-Aldrich

Phenothiazine

SAJ first grade

Synonym(s):

10H-Phenothiazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H9NS
CAS Number:
Molecular Weight:
199.27
Beilstein/REAXYS Number:
143237
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
grade:
SAJ first grade
bp:
371 °C (lit.)
Pricing and availability is not currently available.

grade

SAJ first grade

availability

available only in Japan

dilution

(for analytical testing)

bp

371 °C (lit.)

mp

182-187 °C (lit.)

SMILES string

N1c2ccccc2Sc3ccccc13

InChI

1S/C12H9NS/c1-3-7-11-9(5-1)13-10-6-2-4-8-12(10)14-11/h1-8,13H

InChI key

WJFKNYWRSNBZNX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1 - STOT RE 2 Oral

target_organs

Blood

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maike J Ohlow et al.
Drug discovery today, 16(3-4), 119-131 (2011-01-18)
Rooted in the early days of organic dye chemistry, the phenothiazine structure and its derivatives have since held a prominent place in pharmacology and biomedicine. Initially used for histochemical stains of plasmodia by Paul Ehrlich, anthelmintic and antibiotic properties of
Alina N Sekretaryova et al.
Analytical chemistry, 84(3), 1220-1223 (2011-12-31)
Wiring glucose oxidase in the membrane with an immobilized mediator is possible due to the diffusion ability of the latter, if the enzyme containing membrane is formed according to the proposed protocol, including exposing proteins to water-organic mixtures with the
M L Buck et al.
DICP : the annals of pharmacotherapy, 25(3), 244-247 (1991-03-01)
A two-month-old white girl presented to our facility with increasing lethargy and new onset apnea and bradycardia following a week of upper respiratory tract infection symptoms. The patient had been receiving a cough syrup containing promethazine hydrochloride during the previous
Gabriela B Rodrigues et al.
Journal of photochemistry and photobiology. B, Biology, 116, 89-94 (2012-09-18)
Photodynamic antimicrobial chemotherapy (PACT) is a promising alternative to conventional chemotherapy that can be used to treat localized mycosis. The development of PACT depends on identifying effective and selective PS for the different pathogenic species. The in vitro susceptibilities of
Caiwen Ou et al.
Chemical communications (Cambridge, England), 49(18), 1853-1855 (2013-01-31)
We report a 'super-gelator' of a short peptide capped with phenothiazine acetic acid.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service