Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

24-1160

Sigma-Aldrich

Petrol

JIS special grade, ≥90.0%

Synonym(s):

Gasoline, Petroleum benzin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
15101506
grade:
JIS special grade
assay:
≥90.0%

grade

JIS special grade

assay

≥90.0%

form

liquid

availability

available only in Japan

dilution

(for analytical testing)

InChI

1S/C7H7.BrH.Mg/c1-7-5-3-2-4-6-7;;/h3-6H,1H3;1H;/q;;+1/p-1

InChI key

ZRJNGFJIBZKXTP-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Inhalation - STOT SE 3

target_organs

Central nervous system, Nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

-40.0 °F

flash_point_c

-40 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sujith Prasad W Kumarasinghe et al.
International journal of dermatology, 41(12), 877-880 (2002-12-21)
Many methods have been employed, with variable success, in the treatment of cutaneous myiasis caused by Chrysomya species. Experiment 1: to assess the larvicidal effect of mineral turpentine (MT) and the main ingredient of MT, low aromatic white spirits (LAWS)
S P Lund et al.
Neurotoxicology and teratology, 18(1), 67-76 (1996-01-01)
Exposure for 6 h per day, 5 days per week, during a period of 6 months to the organic solvent dearomatized white spirit (0, 400, and 800 ppm) was studied in rats that were 3 months old when the repeated
White spirit--paint thinner, skin stripper.
I P Mackie et al.
Burns : journal of the International Society for Burn Injuries, 30(1), 86-87 (2003-12-25)
[Attempted suicide by intrapulmonary self-injection of White spirit®].
A Godard et al.
Annales francaises d'anesthesie et de reanimation, 31(12), 975-977 (2012-11-20)
I P Dick et al.
Human & experimental toxicology, 16(11), 652-657 (1998-01-14)
1. The absorption of lindane through human skin was assessed in vitro using static diffusion cells with 50% aqueous ethanol as the receptor fluid. Four formulations of lindane were applied, three of which were commercial preparations. The dermal distribution of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service