Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

24-0390

Sigma-Aldrich

Paraffin wax

SAJ first grade, mp 54-56 °C

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41121804
grade:
SAJ first grade
Pricing and availability is not currently available.

grade

SAJ first grade

form

solid

availability

available only in Japan

dilution

(for analytical testing)

mp

54-56 °C

InChI key

JWHAUXFOSRPERK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

nwg

flash_point_f

602.6 °F - Pensky-Martens closed cup

flash_point_c

317 °C - Pensky-Martens closed cup

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xue-Tong Wang et al.
Environmental science and pollution research international, 20(7), 4742-4749 (2013-01-08)
Short-chain chlorinated paraffins (SCCPs) are extremely complex technical mixtures of polychlorinated n-alkanes with carbon chain lengths from C10 to C13 and chlorine content between 49 and 70%. SCCPs are under consideration for inclusion in the Stockholm Convention on persistent organic
Banu Dilek et al.
Archives of physical medicine and rehabilitation, 94(4), 642-649 (2012-11-29)
To evaluate the efficacy of paraffin bath therapy on pain, function, and muscle strength in patients with hand osteoarthritis. Prospective single-blinded randomized controlled trial. Department of physical medicine and rehabilitation in a university hospital. Patients with bilateral hand osteoarthritis (N=56).
Lindy Rauscher et al.
Journal of hazardous materials, 241-242, 441-449 (2012-10-24)
Surface waters impacted by urban runoff in metropolitan areas are becoming increasingly contaminated with polycyclic aromatic hydrocarbons (PAHs). Slow-release oxidant candles (paraffin-KMnO(4)) are a relatively new technology being used to treat contaminated groundwater and could potentially be used to treat
Laiguo Chen et al.
Journal of chromatography. A, 1274, 36-43 (2013-01-05)
Accurately quantifying short chain chlorinated paraffins (SCCPs) in soil samples with gas chromatograph coupled with electron capture negative ionization mass spectrometry (GC-ECNI-MS) is difficult because many other polychlorinated pollutants are present in the sample matrices. These pollutants (e.g., polychlorinated biphenyls
Zhi-Yong Zhang et al.
Journal of hazardous materials, 243, 105-111 (2012-10-31)
In this study, nanoscale zero-valent iron (NZVI) particles were synthesized and used for the reductive dehalogenation of short chain chlorinated paraffins (SCCPs) in the laboratory. The results show that the dechlorination rate of chlorinated n-decane (CP(10)) by NZVI increased with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service