Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

24-0350

Sigma-Aldrich

Paraffin wax

SAJ first grade, mp 46-48 °C

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41121813
grade:
SAJ first grade
Pricing and availability is not currently available.

grade

SAJ first grade

form

solid

availability

available only in Japan

dilution

(for analytical testing)

mp

46-48 °C

InChI key

JWHAUXFOSRPERK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

13 - Non Combustible Solids

wgk_germany

nwg

flash_point_f

602.6 °F - Pensky-Martens closed cup

flash_point_c

317 °C - Pensky-Martens closed cup

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu Seob Shin et al.
Urology, 81(2), 437-441 (2013-02-05)
To introduce a new repair technique, inverted V-shape, for penile paraffinoma without necrosis of the ventral skin anastomosed. From March 2006 to July 2012, 34 patients underwent penile paraffinoma repair using the T-style or a new operation procedure, the inverted
Wisam Al-Faqheri et al.
PloS one, 8(3), e58523-e58523 (2013-03-19)
This paper introduces novel vacuum/compression valves (VCVs) utilizing paraffin wax. A VCV is implemented by sealing the venting channel/hole with wax plugs (for normally-closed valve), or to be sealed by wax (for normally-open valve), and is activated by localized heating
Roman Zimmermann et al.
Plant physiology, 152(1), 356-365 (2009-11-21)
In an approach to study lateral root development in monocots, genome-wide searches for homologs of the Gibberellic Acid Stimulated Transcript-like (GAST-like) gene family in rice (Oryza sativa) and maize (Zea mays) were carried out. Six novel GAST-like genes in rice
Sharoni Jacobs
Cold Spring Harbor protocols, 2012(11), 1195-1202 (2012-11-03)
The Whole Genome Sampling Analysis (WGSA) assay in combination with Affymetrix GeneChip Mapping Arrays is used for copy number analysis of high-quality DNA samples (i.e., samples that have been collected from blood, fresh or frozen tissue, or cell lines). Formalin-fixed
Abdulazeez Salawu et al.
PloS one, 7(11), e50415-e50415 (2012-12-05)
Most soft tissue sarcomas are characterized by genetic instability and frequent genomic copy number aberrations that are not subtype-specific. Oligonucleotide microarray-based Comparative Genomic Hybridisation (array CGH) is an important technique used to map genome-wide copy number aberrations, but the traditional

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service