Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

23-0380

Sigma-Aldrich

Lemon oil

CP

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
Pricing and availability is not currently available.

grade

CP

availability

available only in Japan

refractive index

n20/D 1.473 (lit.)

bp

176 °C (lit.)

density

0.850 g/mL at 25 °C

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. IS this a natural flavour

    1 answer
    1. Unfortunately, this product has been discontinued.

      Please see the link below to review alternative Lemon Oil products:
      https://www.sigmaaldrich.com/search/8008-56-8?focus=products&page=1&perpage=30&sort=relevance&term=8008-56-8&type=cas_number

      Should documentation be required, please navigate to the link https://www.sigmaaldrich.com/techservice where our Technical Service team can assist you further. We kindly ask you to navigate to the link and click on "Product Documentation" under the Products Section with all the required information so that a member of our team can reach out to you to assist further. Thank you.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service