Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

23-0270

Sigma-Aldrich

Cedar oil

SAJ first grade

Synonym(s):

Cedarwood oil

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
Pricing and availability is not currently available.

grade

SAJ first grade

form

oil

availability

available only in Japan

density

0.952 g/mL at 25 °C (lit.)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Cedar oil demonstrates antimicrobial activity against anaerobic bacteria and yeast, as assessed by a standardized broth assay [1].

signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Skin Irrit. 2 - Skin Sens. 1B

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

183.2 °F

flash_point_c

84 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service