Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

21-3277

Sigma-Aldrich

Nonidet P-40

CP

Synonym(s):

IGEPAL® CA-630, Octylphenoxy poly(ethyleneoxy)ethanol, branched

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H4O)nC14H22O
CAS Number:
MDL number:
UNSPSC Code:
12162002
Pricing and availability is not currently available.

grade

CP

availability

available only in Japan

CMC

0.08 mM (20-25°C)

HLB

13

SMILES string

O(CCO)c1ccc(cc1)C(CC(C)(C)C)(C)C

InChI

1S/C16H26O2/c1-15(2,3)12-16(4,5)13-6-8-14(9-7-13)18-11-10-17/h6-9,17H,10-12H2,1-5H3

InChI key

JYCQQPHGFMYQCF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Nonionic, non-denaturing detergent

Quality

Chemically indistinguishable from Nonidet P-40, which is no longer commercially available.

Caution

May develop turbidity or sediment during storage. A clear liquid can be obtained on heating to 40°C.

Legal Information

IGEPAL is a registered trademark of Solvay
Nonidet is a trademark of Shell Chemical Co.

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

483.8 °F - closed cup

flash_point_c

251 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Michael P Raynor et al.
Journal of hematology & oncology, 2, 24-24 (2009-06-09)
The ability to screen blood of early stage operable breast cancer patients for circulating tumour cells is of potential importance for identifying patients at risk of developing distant relapse. We present the results of a study of the efficacy of
Santhi Gorantla et al.
Glia, 56(2), 223-232 (2007-11-30)
Virus-infected and immune-competent mononuclear phagocytes (MP; perivascular macrophages and microglia) drive the neuropathogenesis of human immunodeficiency virus type 1 (HIV-1) infection. Modulation of the MP phenotype from neurodestructive to neuroprotective underlies adjunctive therapeutic strategies for human disease. We reasoned that
Russell M Morphew et al.
Veterinary parasitology, 191(3-4), 379-385 (2012-10-02)
Fasciola hepatica NP-40 released protein extract (FhNPE) exhibits potent Th1 immunosuppressive properties in vitro and in vivo. However, the protein composition of this active fraction, responsible for Th1 immune modulatory activity, has yet to be resolved. Therefore, FhNPE, a Nonidet
Anita Szodorai et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 29(46), 14534-14544 (2009-11-20)
The amyloid precursor protein (APP) is anterogradely transported by conventional kinesin in a distinct transport vesicle, but both the biochemical composition of such a vesicle and the specific kinesin-1 motor responsible for transport are poorly defined. APP may be sequentially
Halil İbrahim Ulusoy et al.
Talanta, 88, 516-523 (2012-01-24)
A new micelle-mediated separation and preconcentration method was developed for ultra-trace quantities of mercury ions prior to spectrophotometric determination. The method is based on cloud point extraction (CPE) of Hg(II) ions with polyethylene glycol tert-octylphenyl ether (Triton X-114) in the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service