Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

21-3120

Sigma-Aldrich

1-Nitroso-2-naphthol

SAJ special grade, ≥97.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ONC10H6OH
CAS Number:
Molecular Weight:
173.17
Colour Index Number:
10005
Beilstein/REAXYS Number:
776947
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
grade:
SAJ special grade
assay:
≥97.0%
Pricing and availability is not currently available.

grade

SAJ special grade

assay

≥97.0%

form

solid

availability

available only in Japan

dilution

(for analytical testing)

mp

106-108 °C (dec.) (lit.)

SMILES string

Oc1ccc2ccccc2c1N=O

InChI

1S/C10H7NO2/c12-9-6-5-7-3-1-2-4-8(7)10(9)11-13/h1-6,12H

InChI key

YXAOOTNFFAQIPZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[A modified method of determination of bound 5-hydroxyindoleacetic acid in the urine].
N G Shafranova et al.
Laboratornoe delo, (7)(7), 25-26 (1983-01-01)
Nayyef Aljaar et al.
The Journal of organic chemistry, 78(1), 154-166 (2012-11-28)
Reactions between 1-nitroso-2-naphthols and α-functionalized ketones such as α-bromo-, α-chloro-, α-mesyloxy-, α-tosyloxy-, and α-hydroxy ketones under basic conditions delivered 2-substituted naphtho[1,2-d][1,3]oxazoles in a single synthetic operation. The product formation was accompanied by the unexpected loss of the C═O group from
C A Georgiou et al.
The Analyst, 115(3), 309-313 (1990-03-01)
The reaction of para- and meta-substituted phenols with 1-nitroso-2-naphthol in the presence of either CeIV or PbIV as an oxidant has been used to develop a fast automated flow injection (Fl) method. A stopped-flow kinetic study of the reaction revealed
N Takahashi et al.
Free radical research communications, 4(6), 351-358 (1988-01-01)
The nonenzymatic reduction of nitrosobenzene (I), 2-nitroso-1-naphthol (II) and 2-nitroso-1-naphthol-4-sulfonic acid (III) with reducing agents such as NADPH, L-cysteine and N-acetyl-L-cysteine led to the formation of the corresponding hydronitroxide radicals, as confirmed with ESR spectroscopy. In addition to these radicals
C F Burrows et al.
Journal of the American Veterinary Medical Association, 183(3), 318-322 (1983-08-01)
The nitrosonapthol test, which qualitatively measures urinary excretion of 4-hydroxyphenylacetic acid and related compounds, was evaluated as a screening test for small intestinal disease in 60 dogs with chronic diarrhea. Test results were positive in 8 of 13 dogs with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service