Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

21-2700

Sigma-Aldrich

4-Nitrophenol

JIS special grade, ≥99.0%

Synonym(s):

p-Nitrophenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4OH
CAS Number:
Molecular Weight:
139.11
Beilstein/REAXYS Number:
1281877
EC Number:
MDL number:
UNSPSC Code:
12352102
PubChem Substance ID:
grade:
JIS special grade
assay:
≥99.0%
bp:
279 °C (lit.)
vapor pressure:
0.6 mmHg ( 120 °C)
Pricing and availability is not currently available.

grade

JIS special grade

vapor pressure

0.6 mmHg ( 120 °C)

assay

≥99.0%

form

solid

autoignition temp.

541 °F

availability

available only in Japan

dilution

(for analytical testing)

bp

279 °C (lit.)

mp

110-115 °C (lit.)

SMILES string

O=N(C1=CC=C(O)C=C1)=O

InChI

1S/C6H5NO3/c8-6-3-1-5(2-4-6)7(9)10/h1-4,8H

InChI key

BTJIUGUIPKRLHP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - STOT RE 2 Oral

target_organs

Kidney,Liver

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 2

flash_point_f

336.2 °F - closed cup

flash_point_c

169.0 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Azrina Md Ralib et al.
Critical care (London, England), 18(6), 601-601 (2014-11-05)
Acute Kidney Injury (AKI) biomarker utility depends on sample timing after the onset of renal injury. We compared biomarker performance on arrival in the emergency department (ED) with subsequent performance in the intensive care unit (ICU). Urinary and plasma Neutrophil
Siew Hong Lam et al.
Biochimica et biophysica acta, 1830(10), 4778-4789 (2013-06-25)
4-Nitrophenol (4-NP) is a prioritized environmental pollutant and its toxicity has been investigated using zebrafish, advocated as an alternative toxicological model. However, molecular information of 4-NP induced hepatotoxicity is still limited. This study aimed to obtain molecular insights into 4-NP-induced
Jau-Rung Chiou et al.
Journal of hazardous materials, 248-249, 394-400 (2013-02-19)
Silver/iron oxide composite nanoparticles have been synthesized successfully via a facile one-pot green route by the use of l-arginine, which created an aqueous solution of about pH 10 and acted as a reducing agent for the successive formation of iron
Ipsita K Sen et al.
Carbohydrate polymers, 91(2), 518-528 (2012-11-06)
Gold nanoparticles were synthesized by reducing chloroauric acid with a glucan, isolated from an edible mushroom Pleurotus florida, cultivar Assam Florida. Here, glucan acts as reducing as well as stabilizing agent. The synthesized gold nanoparticles were characterized by UV-visible spectroscopy
Ali Mehdinia et al.
Journal of chromatography. A, 1283, 82-88 (2013-03-08)
In this study, an analytical procedure for the selective extraction and detection of 4-nitrophenol (4-NP) was investigated by using of molecularly imprinted polymer on the surface of magnetic nanoparticles (MNPs). The magnetic nanoparticles were modified by tetraethyl orthosilicate (TEOS) and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service