Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

21-0210

Sigma-Aldrich

2-Naphthol

SAJ first grade, ≥98.0%

Synonym(s):

β-Naphthol, 2-Hydroxynaphthalene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7OH
CAS Number:
Molecular Weight:
144.17
Beilstein/REAXYS Number:
742134
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
Pricing and availability is not currently available.

grade

SAJ first grade

vapor density

4.97 (vs air)

vapor pressure

10 mmHg ( 145.5 °C)

assay

≥98.0%

form

solid

availability

available only in Japan

bp

285-286 °C (lit.)

mp

120-122 °C (lit.)

SMILES string

Oc1ccc2ccccc2c1

InChI

1S/C10H8O/c11-10-6-5-8-3-1-2-4-9(8)7-10/h1-7,11H

InChI key

JWAZRIHNYRIHIV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Dam. 1 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

307.4 °F - closed cup

flash_point_c

153 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Supriti Sen et al.
The Analyst, 137(17), 3975-3981 (2012-07-13)
An efficient water soluble fluorescent Al(3+) receptor, 1-[[(2-furanylmethyl)imino]methyl]-2-naphthol (1-H) was synthesized and characterized by physico-chemical and spectroscopic tools along with single crystal X-ray crystallography. High selectivity and affinity of 1-H towards Al(3+) in HEPES buffer (DMSO/water: 1/100) of pH 7.4
Erin E Podlesny et al.
Organic letters, 14(6), 1408-1411 (2012-03-01)
The first enantioselective total synthesis of the bisanthraquinone (S)-bisoranjidiol and an unnatural regioisomer has been accomplished. Key features of the synthesis include the asymmetric oxidative biaryl coupling of a hindered 8-substituted 2-naphthol, selective para-quinone formation, and regioselective tandem Diels-Alder/aromatization reactions.
Glenn Talaska et al.
Toxicology letters, 213(1), 45-48 (2011-05-26)
Urinary bladder cancer is a historical disease of rubber workers often been associated with exposure to aromatic amines such as 2-naphthylamine. While exposure to these compounds has decreased markedly over time, the bladder cancer risk has not decreased in direct
L E King et al.
Environmental science & technology, 48(19), 11637-11645 (2014-08-26)
Hamilton Harbour, Ontario, Canada is one of the most polluted sites on the Great Lakes, and is subject to substantial airborne pollution due to emissions from both heavy industry and intense vehicle traffic. Mutagenic Polycyclic aromatic hydrocarbons (PAHs) are present
H Kim et al.
European journal of clinical nutrition, 65(10), 1118-1125 (2011-05-19)
Exposure to polycyclic aromatic hydrocarbons (PAHs) during pregnancy is known to increase oxidative stress, which may influence pregnancy outcomes and health of the child. This study investigated whether fruit and vegetable intake modifies the relationship between exposure to PAHs and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service