Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

19-4543

Sigma-Aldrich

Molecular sieves, 3 Å

CP, 1/8 diameter

Synonym(s):

Molecular Sieve UOP Type 3Å

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KnNa12-n[(AlO2)12(SiO2)12] · xH2O
CAS Number:
MDL number:
UNSPSC Code:
23201100
Pricing and availability is not currently available.

grade

CP

form

solid

availability

available only in Japan

diameter

1/8

pore size

3 Å

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service