Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

19-2900

Sigma-Aldrich

Methyl cellulose

CP, viscosity 400 cP 

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
Pricing and availability is not currently available.

grade

CP

availability

available only in Japan

viscosity

400 cP

SMILES string

[*]OC[C@H]1O[C@@H](O[C@@H]2[C@@H](CO[*])O[C@@H](O[*])[C@H](O[*])[C@H]2O[*])[C@H](O[*])[C@@H](O[*])[C@@H]1O[*]

InChI key

YLGXILFCIXHCMC-JHGZEJCSSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elsa-Noah N'Diaye et al.
EMBO reports, 10(2), 173-179 (2009-01-17)
Ubiquilins (UBQLNs) are adaptor proteins thought to deliver ubiquitinated substrates to proteasomes. Here, we show a role for UBQLN in autophagy: enforced expression of UBQLN protects cells from starvation-induced death, whereas depletion of UBQLN renders cells more susceptible. The UBQLN
Matthew J Caicco et al.
Journal of controlled release : official journal of the Controlled Release Society, 166(3), 197-202 (2013-01-12)
Stimulation of endogenous neural stem/progenitor cells (NSPCs) with therapeutic factors holds potential for the treatment of stroke. Cyclosporin A (CsA) is a particularly promising candidate molecule because it has been shown to act as a survival factor for these cells
K S Sink et al.
Behavioural brain research, 243, 286-293 (2013-02-05)
Calcitonin gene-related peptide (CGRP) acting within the bed nucleus of the stria terminalis (BNST) increases anxiety as well as neural activation in anxiety-related structures, and mediates behavioral stress responses. Similar effects have been described following intra-ventricular as well as intra-BNST
Yihua Wang et al.
Biochemistry, 52(9), 1611-1621 (2013-02-07)
Myosin powers contraction in heart and skeletal muscle and is a leading target for mutations implicated in inheritable muscle diseases. During contraction, myosin transduces ATP free energy into the work of muscle shortening against resisting force. Muscle shortening involves relative
Walter Fuchs et al.
Journal of virology, 83(8), 3930-3943 (2009-02-06)
Cleavage and encapsidation of newly replicated herpes simplex virus type 1 (HSV-1) DNA requires several essential viral gene products that are conserved in sequence within the Herpesviridae. However, conservation of function has not been analyzed in greater detail. For functional

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service