Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

19-2350

Sigma-Aldrich

Methyl acetate

SAJ first grade, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOCH3
CAS Number:
Molecular Weight:
74.08
Beilstein/REAXYS Number:
1736662
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
grade:
SAJ first grade
assay:
≥99.0%
bp:
57-58 °C (lit.)
vapor pressure:
165 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

SAJ first grade

vapor density

2.55 (vs air)

vapor pressure

165 mmHg ( 20 °C)

assay

≥99.0%

form

liquid

autoignition temp.

936 °F

expl. lim.

16 %

availability

available only in Japan

dilution

(for analytical testing)

refractive index

n20/D 1.361 (lit.)

bp

57-58 °C (lit.)

mp

−98 °C (lit.)

density

0.934 g/mL at 25 °C

SMILES string

COC(C)=O

InChI

1S/C3H6O2/c1-3(4)5-2/h1-2H3

InChI key

KXKVLQRXCPHEJC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

target_organs

Central nervous system

supp_hazards

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

8.6 °F - closed cup

flash_point_c

-13 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Franc Avbelj et al.
Proceedings of the National Academy of Sciences of the United States of America, 106(9), 3137-3141 (2009-02-10)
Recent calorimetric measurements of the solvation enthalpies of some dipeptide analogs confirm our earlier prediction that the principle of group additivity is not valid for the interaction of the peptide group with water. We examine the consequences for understanding the
Marco Candelaresi et al.
The journal of physical chemistry. A, 113(46), 12783-12790 (2009-10-02)
The solvation dynamics of methyl acetate in heavy water are analyzed by means of two-dimensional infrared spectroscopy, in conjunction with Car-Parrinello molecular dynamics simulations. The C horizontal lineO stretching infrared band of methyl acetate in water splits into a doublet
DeLu Tyler Yin et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 18(26), 8130-8139 (2012-05-24)
Several serine hydrolases catalyze a promiscuous reaction: perhydrolysis of carboxylic acids to form peroxycarboxylic acids. The working hypothesis is that perhydrolases are more selective than esterases for hydrogen peroxide over water. In this study, we tested this hypothesis, and focused
M L Senent et al.
The Journal of chemical physics, 138(4), 044319-044319 (2013-02-08)
Highly correlated ab initio calculations (CCSD(T)) are used to compute gas phase spectroscopic parameters of three isotopologues of the methyl acetate (CH(3)COOCH(3), CD(3)COOCH(3), and CH(3)COOCD(3)), searching to help experimental assignments and astrophysical detections. The molecule shows two conformers cis and
Lei Yang et al.
The journal of physical chemistry. A, 112(28), 6364-6372 (2008-06-21)
The mechanisms and the kinetics of the OH (OD) radicals with methyl acetate CH3C(O)OCH3 are investigated theoretically. The dual-level direct dynamics method is employed in the calculation of the rate constants. The optimized geometries and frequencies and the gradients of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service