Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

19-2080

Sigma-Aldrich

Acetyl-β-methylcholine chloride

SAJ special grade, ≥98.0%

Synonym(s):

(2-Acetoxypropyl)trimethylammonium chloride, Methacholine chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOCH(CH3)CH2N(Cl)(CH3)3
CAS Number:
Molecular Weight:
195.69
Beilstein/REAXYS Number:
4162373
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
grade:
SAJ special grade
assay:
≥98.0%
Pricing and availability is not currently available.

grade

SAJ special grade

assay

≥98.0%

availability

available only in Japan

dilution

(for analytical testing)

mp

171-173 °C (lit.)

SMILES string

[Cl-].CC(C[N+](C)(C)C)OC(C)=O

InChI

1S/C8H18NO2.ClH/c1-7(11-8(2)10)6-9(3,4)5;/h7H,6H2,1-5H3;1H/q+1;/p-1

InChI key

JHPHVAVFUYTVCL-UHFFFAOYSA-M

Gene Information

human ... CHRM3(1131)

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rifat A Chaudry et al.
Thorax, 69(6), 580-585 (2014-02-14)
Asthma and airway hyper-responsiveness are reportedly more common in children with sickle cell disease (SCD). To determine airway responsiveness, airway inflammation and clinical features of asthma in SCD. A prospective, single-centre study of 50 SCD children without overt pulmonary vascular
Nizar Naji et al.
Respiratory medicine, 107(4), 503-510 (2013-01-29)
Lung function tests are essential for the diagnosis and management of bronchial asthma. Impulse oscillation (IOS) system is an alternative way to measure lung mechanics for some patients. We investigated the relative sensitivities of IOS, body plethysmography and spirometry in
Roy H E Cloots et al.
American journal of physiology. Lung cellular and molecular physiology, 305(5), L364-L376 (2013-07-09)
Asthma is a chronic inflammatory disease of the small airways, with airway hyperresponsiveness (AHR) and inflammation as hallmarks. Recent studies suggest a role for arginase in asthma pathogenesis, possibly because arginine is the substrate for both arginase and NO synthase
Mark J Hoegger et al.
Science (New York, N.Y.), 345(6198), 818-822 (2014-08-16)
Lung disease in people with cystic fibrosis (CF) is initiated by defective host defense that predisposes airways to bacterial infection. Advanced CF is characterized by a deficit in mucociliary transport (MCT), a process that traps and propels bacteria out of
Sue R Downie et al.
Journal of applied physiology (Bethesda, Md. : 1985), 114(6), 770-777 (2013-02-02)
The forced oscillation technique (FOT) and multiple-breath nitrogen washout (MBNW) are noninvasive tests that are potentially sensitive to peripheral airways, with MBNW indexes being especially sensitive to heterogeneous changes in ventilation. The objective was to study methacholine-induced changes in the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service