Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

19-1930

Sigma-Aldrich

Mercury(II) thiocyanate

JIS special grade, ≥99.0%

Synonym(s):

Mercuric isothiocyanate, Mercuric sulfocyanide, Mercuric thiocyanate, Mercury di(thiocyanate), Thiocyanic acid, mercury(II) salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Hg(SCN)2
CAS Number:
Molecular Weight:
316.75
Beilstein/REAXYS Number:
3561344
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
assay:
≥99.0%
grade:
JIS special grade
form:
solid
Pricing and availability is not currently available.

grade

JIS special grade

assay

≥99.0%

form

solid

reaction suitability

reagent type: catalyst
core: mercury

availability

available only in Japan

mp

165 °C (dec.) (lit.)

density

3.71 g/mL at 25 °C (lit.)

SMILES string

N#CS[Hg]SC#N

InChI

1S/2CHNS.Hg/c2*2-1-3;/h2*3H;/q;;+2/p-2

InChI key

GBZANUMDJPCQHY-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

signalword

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

supp_hazards

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

248.0 °F - closed cup

flash_point_c

120 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M A Koupparis et al.
Journal of pharmaceutical sciences, 75(8), 800-804 (1986-08-01)
An automated flow-injection determination of chloride salts of drugs, based on the colorimetric mercurothiocyanate determination of chloride counterion is described. The majority of these drugs can be determined, using chloride ion standards, with a coefficient of variation less than 1%
Ajit Kumar et al.
World journal of microbiology & biotechnology, 31(8), 1211-1216 (2015-05-11)
Xanthobacter autotrophicus GJ10 has been widely studied because of its ability to degrade halogenated compounds, especially 1,2-dichloroethane (1,2-DCA), which is achieved through chromosomal as well as plasmid pAUX1 encoded 1,2-DCA degrading genes. This work described the gene expression and enzyme

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service