Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

19-0570

Sigma-Aldrich

Maleic acid

SAJ special grade, ≥99.0%

Synonym(s):

cis-Butenedioic acid, Toxilic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCH=CHCO2H
CAS Number:
Molecular Weight:
116.07
Beilstein/REAXYS Number:
605762
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
Pricing and availability is not currently available.

grade

SAJ special grade

assay

≥99.0%

form

fine crystals
powder

availability

available only in Japan

mp

130-135 °C (lit.)

solubility

water: 478.8 g/L at 20 °C

density

1.59 g/mL at 25 °C (lit.)

SMILES string

OC(=O)\C=C/C(O)=O

InChI

1S/C4H4O4/c5-3(6)1-2-4(7)8/h1-2H,(H,5,6)(H,7,8)/b2-1-

Inchi Key

VZCYOOQTPOCHFL-UPHRSURJSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chih-Chia Huang et al.
ACS applied materials & interfaces, 7(45), 25259-25269 (2015-10-27)
We have successfully introduced a proton-induced controlled reaction of HAuCl4 and poly(styrene-alt-maleic acid) (PSMA) sodium salt to prepare triangular and multicore Au@polymer nanoparticles (NPs). The interparticle interactions in the core gave rise to an absorption band at the near-infrared wavelength.
J M Herreras et al.
Ophthalmology, 99(7), 1082-1088 (1992-07-01)
This study was undertaken to see whether long-term locally applied ocular medications produced any alterations in the ocular surface, and, in particular, whether it caused damage to the mucus layer of the tear film. The authors studied the ocular surface
Hanna Kotikoski et al.
Acta ophthalmologica Scandinavica, 81(4), 355-360 (2003-07-16)
To investigate whether a nitric oxide donor given as a single oral dose is able to modify aqueous humour flow in healthy volunteers. Ten healthy volunteers participated in a randomized, double-masked and placebo-controlled cross-over study. Aqueous humour flow was assessed
K Grundke et al.
Advances in colloid and interface science, 222, 350-376 (2014-12-10)
Contact angle hysteresis phenomena on polymer surfaces have been studied by contact angle measurements using sessile liquid droplets and captive air bubbles in conjunction with a drop shape method known as Axisymmetric Drop Shape Analysis - Profile (ADSA-P). In addition
Clare A Daykin et al.
Analytical biochemistry, 304(2), 220-230 (2002-05-16)
Blood plasma is the major vehicle by which metabolites are transported around the body in mammalian species, and chemical analysis of plasma can provide a wealth of information relating to the biochemical status of an individual and is important for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service