Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

19-0390

Sigma-Aldrich

Magnesium perchlorate

SAJ first grade, ≥80.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Mg(ClO4)2
CAS Number:
Molecular Weight:
223.21
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
assay:
≥80.0%
grade:
SAJ first grade
form:
solid
Pricing and availability is not currently available.

grade

SAJ first grade

assay

≥80.0%

form

solid

reaction suitability

reagent type: oxidant

availability

available only in Japan

SMILES string

[Mg++].[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O

InChI

1S/2ClHO4.Mg/c2*2-1(3,4)5;/h2*(H,2,3,4,5);/q;;+2/p-2

InChI key

MPCRDALPQLDDFX-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

pictograms

Flame over circleExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

5.1A - Strongly oxidizing hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J D Anastassopoulou et al.
Magnesium research, 3(1), 15-18 (1990-03-01)
This study examined the effects on the radiolysis of magnesium-guanosine-5'-monophosphate complexes in deaerated aqueous solutions, investigated by Fourier transform infrared (FT-IR) spectroscopy. It was found that when the system was irradiated with a dose rate of 500 rad/min (low dose
F Javier Céspedes-Guirao et al.
The Journal of organic chemistry, 74(16), 5871-5880 (2009-07-29)
Zinc phthalocyanine-perylenebisimide pentameric arrays, ZnPc(PDI)(4) 1 and 2, have been synthesized. ZnPc(PDI)(4) 1 has no substituents in the PDI bay positions, while ZnPc(PDI)(4) 2 presents four phenoxy groups in the bay positions of each perylene. In both cases, the PDI
J D Anastassopoulou
Magnesium research, 3(1), 19-21 (1990-03-01)
The mechanism of the gamma-radiolysis of the nucleotide guanosine-5'-monophosphate (5'-GMP) in the presence of Mg++ ions and cis-platinum was investigated. From high performance liquid chromatography and spectroscopy studies it was found that the OH radicals, produced in irradiated aqueous solutions
V R Nikolaevskaia et al.
Voprosy pitaniia, (3)(3), 58-60 (1990-05-01)
In experiments on rats it was established that the compound CAM-300 in a dose of 2 mg/kg did not induce body mass growth, protein biological value increase or pepsinogen concentration change in the gastric mucosa. Fenobolin stimulation of growth in
T Theophanides
Magnesium research, 9(4), 259-262 (1996-12-01)
The interaction of Mg2+ cations in biological systems is studied by using nucleic acid bases as the biological system. Magnesium salts, such as, MgCl2 6H2O, MgSO4. 7H2O and Mg(ClO4). XH2O have been employed in order to compare their complexation with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service